Skip to Content
Merck
All Photos(1)

Documents

56218

Supelco

Nystose

analytical standard

Synonym(s):

β-D-Fruf-(2->1)-β-D-Fruf-(2->1)-β-D-Fruf-(2<-->1)-α-D-Glup

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H42O21
CAS Number:
Molecular Weight:
666.58
Beilstein:
6171779
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

analyte chemical class(es)

oligosaccharides

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

OC[C@H]1O[C@H](O[C@]4(CO[C@]3(CO[C@]2(CO)O[C@H](CO)[C@@H](O)[C@@H]2O)O[C@H](CO)[C@@H](O)[C@@H]3O)O[C@H](CO)[C@@H](O)[C@@H]4O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C24H42O21/c25-1-8-12(30)16(34)17(35)21(41-8)45-24(20(38)15(33)11(4-28)44-24)7-40-23(19(37)14(32)10(3-27)43-23)6-39-22(5-29)18(36)13(31)9(2-26)42-22/h8-21,25-38H,1-7H2/t8-,9-,10-,11-,12-,13-,14-,15-,16+,17-,18+,19+,20+,21-,22-,23-,24+/m1/s1

InChI key

FLDFNEBHEXLZRX-DLQNOBSRSA-N

General description

Nystose is one of the smallest insulin related oligosaccharide, which can be enzymatically obtained from sucrose.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

NMR spectroscopy of nystose.
J W Timmermans et al.
Carbohydrate research, 243(2), 379-384 (1993-05-07)
Yijia Xie et al.
AMB Express, 7(1), 128-128 (2017-06-24)
The industrial microorganisms used for fructooligosaccharides (FOS) synthesis are generally fermented with sucrose as carbon source to induce the production of β-fructofuranosidase (FFase) having transfructosylation activity. Consequently, isolation of novel FFase producers from a sucrose-enriched biotope would help improve FOS
A Magri et al.
Journal of applied microbiology, 128(5), 1414-1426 (2020-01-01)
Fructooligosaccharides (FOSs) known for their health properties and β-(2→6)-levan-type FOSs have shown prebiotic and immunomodulatory activities that overcome those of commercial β-(2→1)-FOSs, but costs do not favour their use. Moreover, FOSs can reach the bloodstream through the diet, and little
M Lüscher et al.
FEBS letters, 385(1-2), 39-42 (1996-04-29)
Sucrose-sucrose 1-fructosyltransferase (1-SST) was purified 100-fold from tubers of Helianthus tuberosus L. The purified enzyme was essentially devoid of invertase activity and could be separated by isoelectric focusing into five isoforms which all were composed of two subunits (59 and
M Iacovou et al.
Journal of human nutrition and dietetics : the official journal of the British Dietetic Association, 31(2), 256-265 (2017-06-21)
To investigate if a low fermentable oligo-, di- and mono-saccharides and polyols (FODMAP) diet consumed by breastfeeding mothers may be associated with reduced symptoms of infantile colic. Exclusively breastfeeding mothers and their typically-developing healthy infants who met the Wessel Criteria

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service