Skip to Content
Merck
All Photos(1)

Key Documents

50239

Sigma-Aldrich

Gly-Gly-Gly

≥99.0% (NT), suitable for ligand binding assays, BioUltra

Synonym(s):

Glycyl-glycyl-glycine, Triglycine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2CONHCH2CONHCH2COOH
CAS Number:
Molecular Weight:
189.17
Beilstein:
1711130
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

product name

Gly-Gly-Gly, BioUltra, ≥99.0% (NT)

product line

BioUltra

Quality Level

Assay

≥99.0% (NT)

form

powder

technique(s)

ligand binding assay: suitable

impurities

insoluble matter, passes filter test

ign. residue

≤0.1% (as SO4)

loss

≤0.5% loss on drying, 110 °C

color

white

pH

4.5-6.0 (25 °C, 0.5 M in H2O)

solubility

H2O: 0.5 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤50 mg/kg
Bi: ≤5 mg/kg
Ca: ≤20 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

λ

0.5 M in H2O

UV absorption

λ: 260 nm Amax: 0.15
λ: 280 nm Amax: 0.10

SMILES string

NCC(=O)NCC(=O)NCC(O)=O

InChI

1S/C6H11N3O4/c7-1-4(10)8-2-5(11)9-3-6(12)13/h1-3,7H2,(H,8,10)(H,9,11)(H,12,13)

InChI key

XKUKSGPZAADMRA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Gly-Gly-Gly

General description

Gly-Gly-Gly or triglycine is a flexible peptide linker with three glycine residues linked by peptide bonds in a linear sequence. Gly-linkers link multiple domains in a single protein without affecting the function of each domain.

Application

<ul>
<li><strong>Targeting PEPT1: a novel strategy to improve the antitumor efficacy of doxorubicin in human hepatocellular carcinoma therapy:</strong> Research explores Gly-Gly-Gly peptides as potential enhancers for drug transport efficiency, positioning them as valuable tools in clinical diagnostic peptide development for targeted cancer therapies (Gong et al., 2017).</li>
<li><strong>Iminopyridine complexes of manganese, rhenium, and molybdenum derived from amino ester methylserine and peptides Gly-Gly, Gly-Val, and Gly-Gly-Gly: self-assembly of the peptide chains:</strong> This study demonstrates the use of Gly-Gly-Gly in the synthesis of complex metal-peptide frameworks for potential use in high-purity peptide applications in IVD manufacturing (&Aacute;lvarez et al., 2012).</li>
</ul>

Biochem/physiol Actions

Substrate for reproducible serum protein measurements by the biuret reaction and for the assay of aminotripeptidases.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vishnu Priyanka Reddy Chichili et al.
Protein science : a publication of the Protein Society, 22(2), 153-167 (2012-12-12)
Linkers or spacers are short amino acid sequences created in nature to separate multiple domains in a single protein. Most of them are rigid and function to prohibit unwanted interactions between the discrete domains. However, Gly-rich linkers are flexible, connecting
Sudipta Chakraborty et al.
Bioconjugate chemistry, 21(5), 969-978 (2010-04-15)
This report presents the synthesis and evaluation of (111)In(DOTA-6G-RGD(4)) (DOTA = 1,4,7,10-tetraazacyclododecane-1,4,7,10-tetracetic acid; 6G-RGD(4) = E{G(3)-E[G(3)-c(RGDfK)](2)}(2) and G(3) = Gly-Gly-Gly), (111)In(DOTA-RGD(4)) (RGD(4) = E{E[c(RGDfK)](2)}(2)) and (111)In(DOTA-3G-RGD(2)) (3G-RGD(2) = G(3)-E[G(3)-c(RGDfK)](2)) as new radiotracers for imaging integrin alpha(v)beta(3)-positive tumors. The IC(50) values
Craig P Schwartz et al.
Proceedings of the National Academy of Sciences of the United States of America, 107(32), 14008-14013 (2010-07-28)
Nitrogen K-edge spectra of aqueous triglycine were measured using liquid microjets, and the effects of Hofmeister-active salts on the spectra were observed. Spectra simulated using density functional theory, sampled from room temperature classical molecular dynamics trajectories, capture all major features
Dipositively charged protonated a3 and a2 ions: generation by fragmentation of [La(GGG)(CH3CN)2]3+.
Tujin Shi et al.
Angewandte Chemie (International ed. in English), 47(43), 8288-8291 (2008-09-26)
Avisek Chatterjee et al.
The Journal of chemical physics, 129(10), 105104-105104 (2008-12-03)
X-ray photoelectron spectroscopy (XPS) has been used to investigate the core-level electronic structures of glycine (G) and its peptides, including glycyl-glycine (GG), diglycyl-glycine (GGG), and polyglycine (poly-G), in their powder forms. Increasing the number of G units in the peptides

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service