Skip to Content
Merck
All Photos(5)

Documents

C8754

Sigma-Aldrich

Thiamine pyrophosphate

≥95%, powder

Synonym(s):

Aneurinepyrophosphoric acid, Cocarboxylase, Thiamine pyrophosphate chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H19ClN4O7P2S
CAS Number:
Molecular Weight:
460.77
Beilstein:
3875902
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.54

product name

Thiamine pyrophosphate, ≥95%

Assay

≥95%

form

powder

solubility

H2O: soluble 50 mg/mL, clear to very slightly hazy, colorless

storage temp.

−20°C

SMILES string

CC1=C(CCOP(OP(O)(O)=O)(O)=O)SC=[N+]1CC2=C(N)N=C(C)N=C2.[Cl-]

InChI

1S/C12H18N4O7P2S.ClH/c1-8-11(3-4-22-25(20,21)23-24(17,18)19)26-7-16(8)6-10-5-14-9(2)15-12(10)13;/h5,7H,3-4,6H2,1-2H3,(H4-,13,14,15,17,18,19,20,21);1H

InChI key

YXVCLPJQTZXJLH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Thiamine pyrophosphate from Sigma has been used as a substrate to assay the enzymes in golgi lamellae isolated from rat tissue.
Thiamine pyrophosphate has been used:
  • for the preparation of chitosan nano-particles
  • as a standard for vitamin B1 and its derivative in high-performance
  • liquid chromatography (HPLC) analysis
  • immunofluorescence

Biochem/physiol Actions

Thiamine pyrophosphate (TPP) is a coenzyme of transketolase, that catalyzes the cleavage of ribulose-5-phosphate. This reaction leads to the formation of D-glyceraldehyde-3-phosphate. It requires the addition of an acceptor aldehyde such as ribose-5-phosphate, glyceraldehye or glycolaldehyde. TPP is capable of decarboxylating hydroxypyruvate in the presence of an ′acceptor aldehyde′.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thiamine pyrophosphate, a coenzyme of transketolase
Racker E, et al.
Journal of the American Chemical Society, 75(4), 1010-1011 (1953)
Nucleosidediphosphatase activity in the Golgi apparatus and its usefulness for cytological studies.
A B NOVIKOFF et al.
Proceedings of the National Academy of Sciences of the United States of America, 47, 802-810 (1961-06-15)
Michael Moulin et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(36), 14622-14627 (2013-08-21)
Thiamin (vitamin B1) is an essential micronutrient needed as a cofactor for many central metabolic enzymes. Animals must have thiamin in their diet, whereas bacteria, fungi, and plants can biosynthesize it de novo from the condensation of a thiazole and
Effects of anti-CD44 monoclonal antibody IM7 carried with chitosan polylactic acid-coated nano-particles on the treatment of ovarian cancer
Yang Y, et al.
Oncology Letters, 13(1), 99-104 (2017)
Enhancement of thiamine release during synthetic mutualism between Chlorella sorokiniana and Azospirillum brasilense growing under stress conditions
Palacios OA, et al.
Journal of Applied Phycology, 28(3), 1521-1531 (2016)

Articles

Explore thiamine's impact on serum-free cell culture systems for biomanufacturing monoclonal antibodies.

Explore thiamine's impact on serum-free cell culture systems for biomanufacturing monoclonal antibodies.

Explore thiamine's impact on serum-free cell culture systems for biomanufacturing monoclonal antibodies.

Explore thiamine's impact on serum-free cell culture systems for biomanufacturing monoclonal antibodies.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service