Skip to Content
Merck
All Photos(1)

Key Documents

F8150

Sigma-Aldrich

D-(+)-Fucose

≥97% (GC)

Synonym(s):

6-Deoxy-D-galactose, Rhodeose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O5
CAS Number:
Molecular Weight:
164.16
Beilstein:
1723320
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

Assay

≥97% (GC)

form

powder

optical activity

[α]20/D 74 to 76 °, c = 4% (w/v) in water

technique(s)

gas chromatography (GC): suitable

color

white

mp

144-145 °C (lit.)

solubility

H2O: 100 mg/mL, clear to slightly hazy, colorless to faintly yellow

storage temp.

room temp

SMILES string

C[C@@H](O)[C@H](O)[C@H](O)[C@@H](O)C=O

InChI

1S/C6H12O5/c1-3(8)5(10)6(11)4(9)2-7/h2-6,8-11H,1H3/t3-,4+,5+,6-/m1/s1

InChI key

PNNNRSAQSRJVSB-DPYQTVNSSA-N

Looking for similar products? Visit Product Comparison Guide

Application

D-Fucose (6-deoxy-D-galactose, rhodeose) may be used to study enzymes involved in its mutarotation to form L-fucose.[1] It may be used as a reference compound in the analysis of carbohydrate metabolites. D-Fucose may be used to study biological processes such as adhesion.[2]

Biochem/physiol Actions

D-Fucose is a nonmetabolizable analog of L-Arabinose.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nancy G Isern et al.
Biotechnology for biofuels, 6(1), 47-47 (2013-04-05)
Caldicellulosiruptor saccharolyticus is a thermophilic, Gram-positive, non-spore forming, strictly anaerobic bacterium of interest in potential industrial applications, including the production of biofuels such as hydrogen or ethanol from lignocellulosic biomass through fermentation. High-resolution, solution-state nuclear magnetic resonance (NMR) spectroscopy is
Daniel J Becker et al.
Glycobiology, 13(7), 41R-53R (2003-03-26)
Fucose is a deoxyhexose that is present in a wide variety of organisms. In mammals, fucose-containing glycans have important roles in blood transfusion reactions, selectin-mediated leukocyte-endothelial adhesion, host-microbe interactions, and numerous ontogenic events, including signaling events by the Notch receptor
Sarah Meinhardt et al.
Nucleic acids research, 40(21), 11139-11154 (2012-09-12)
LacI/GalR transcription regulators have extensive, non-conserved interfaces between their regulatory domains and the 18 amino acids that serve as 'linkers' to their DNA-binding domains. These non-conserved interfaces might contribute to functional differences between paralogs. Previously, two chimeras created by domain
Xiaonan Zhou et al.
PloS one, 13(10), e0204152-e0204152 (2018-10-09)
Lonicera japonica is a typical Chinese herbal medicine. We previously reported a method to isolate polysaccharides from Lonicera japonica (LJP). In this study, we first performed a qualitative analysis of LJP using the Fourier Transform Infrared Spectrometer (FT-IR) and explored
Hiroaki Masuzaki et al.
Applied biochemistry and biotechnology, 181(1), 65-82 (2016-08-11)
Two novel sugar acid-binding lectins were purified from Haplomitrium mnioides (Lindb.) Schust. using a procedure consisting of ammonium sulfate precipitation, G-50 gel filtration, hydroxyapatite chromatography, and HW-50 gel filtration. We reported their partial physicochemical properties: molecular weight, affinity for carbohydrates

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service