Skip to Content
Merck
All Photos(1)

Documents

63580

Millipore

D-(+)-Mannose

≥99%, suitable for microbiology

Synonym(s):

D-Mannopyranose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
Beilstein:
1564373
EC Number:
MDL number:
UNSPSC Code:
41106212
PubChem Substance ID:
NACRES:
NA.85

Quality Level

Assay

≥99%
≥99.0% (sum of enantiomers, HPLC)

form

powder

optical activity

[α]20/D +13.8±0.5°, 24 hr, c = 10% in H2O

ign. residue

≤0.1% (as SO4)

loss

≤0.2% loss on drying, 20 °C (HV)

mp

133-140 °C (lit.)

solubility

H2O: 0.1 g/mL, clear, colorless

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

As: ≤0.1 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

application(s)

microbiology

SMILES string

OC[C@H]1OC(O)[C@@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2/t2-,3-,4+,5+,6?/m1/s1

InChI key

WQZGKKKJIJFFOK-QTVWNMPRSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Otter et al.
Hepatology (Baltimore, Md.), 16(1), 54-59 (1992-07-01)
Various studies have shown that mannose receptors rapidly eliminate glycoproteins and microorganisms bearing high mannose-type carbohydrate chains from the blood circulation. The purpose of this study was to characterize the mannose receptor in the liver, which in vivo is involved
Xiyan Wang et al.
Marine drugs, 19(2) (2021-03-07)
Genome mining of pigmented Pseudoalteromonas has revealed a large potential for the production of bioactive compounds and hydrolytic enzymes. The purpose of the present study was to explore this bioactivity potential in a potent antibiotic and enzyme producer, Pseudoalteromonas rubra
I Adlerberth et al.
Applied and environmental microbiology, 62(7), 2244-2251 (1996-07-01)
Two Lactobacillus plantarum strains of human intestinal origin, strains 299 (= DSM 6595) and 299v (= DSM 9843), have proved to be efficient colonizers of the human intestine under experimental conditions. These strains and 17 other L. plantarum strains were
Chia-Suei Hung et al.
Molecular microbiology, 44(4), 903-915 (2002-05-16)
The first step in the colonization of the human urinary tract by pathogenic Escherichia coli is the mannose-sensitive binding of FimH, the adhesin present at the tip of type 1 pili, to the bladder epithelium. We elucidated crystallographically the interactions
A T Van der Ploeg et al.
The Journal of clinical investigation, 87(2), 513-518 (1991-02-01)
The lysosomal storage disorder glycogenosis type II is caused by acid alpha-glucosidase deficiency. In this study we have investigated the possible applicability of mannose 6-phosphate receptor-mediated enzyme replacement therapy to correct the enzyme deficiency in the most affected tissues. Bovine

Articles

Culture media provides a habitat with suitable nutrients, energy sources, and certain environmental conditions for the growth of microorganisms. The components of the culture media range from simple sugars to peptones, salts, antibiotics, and complex indicators.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service