Skip to Content
Merck

F5262

Anti-Mouse IgG (Fab specific)−FITC antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Goat Anti-Mouse IgG (Fab specific)−Fluorescein isothiocyanate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46
MDL number:
Conjugate:
FITC conjugate
Clone:
polyclonal
Application:
IF, IHC (p)
Citations:
48
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

goat

Quality Level

conjugate

FITC conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

storage condition

protect from light

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:150, indirect immunofluorescence: 1:128

storage temp.

−20°C

target post-translational modification

unmodified

General description

IgG antibody subtype is the most abundant serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids and provides protection from infections caused by bacteria, fungi and viruses. Maternal IgG is transferred to fetus through the placenta that is vital for immune defence of the neonate against infections. Anti-Mouse IgG (Fab specific)-FITC antibody is specific for Fab fragment of all mouse IgG subclasses. It shows no reactivity with mouse IgG, Fc fragment, or human IgG. Goat anti-Mouse IgG is conjugated to FITC and then purified by gel filtration to remove free FITC. The antibody is adsorbed with human IgG to ensure minimal cross reactivity.
Immunoglobulins (Igs) belong to the immunoglobulin super-family. The four classes of IgG include IgG1, IgG2, IgG3 and IgG4.. The IgG heavy chain region is mapped to human chromosome 14. The immunoglobulin structure comprises of two heavy (H) and two light (L) chains, held together by disulfide linkages. The heavy chain has one variable N-terminal region and three to four constant (CH1-CH4) C-terminal regions. The L chain comprises of one variable N-terminal region and a constant C-terminal region.
Mouse IgG is a plasma B cell derived antibody isotype defined by its heavy chain. IgG is the most abundant antibody isotype found in mouse serum. IgG crosses the placental barrier, is a complement activator and binds to the Fc-receptors on phagocytic cells. The level of IgG may vary with the status of disease or infection.
Fluorescein isothiocyanate (FITC) is a fluorescein derivative (fluorochrome) used to tag antibodies, including secondary antibodies, for use in fluorescence-based assays and procedures. FITC excites at 495 nm and emits at 521 nm.

Immunogen

Purified mouse IgG, Fab fragment

Application

Anti-Mouse IgG (Fab specific) FITC antibody produced in goat has been used in:
  • immunolocalization of pancreases or isolated islets
  • immunocytochemistry analysis human tumor cell lines
  • immunohistochemistry of articular cartilage secondary antibody

Anti-Mouse IgG (Fab specific)-FITC antibody may be used for immunofluorescence of human peripheral blood lymphocytes at a minimum dilution of 1:128. It may be used for imunohistochemistry of formalin-fixed, paraffin-embedded human tonsil slides at a working antibody dilution of 1:150. In an agar diffusion assay the conjugate produces a precipitation arc at a dilution of 1:32 versus a 1:80 dilution of normal mouse serum. The antibody was used at a dilution range of 1:50 to 1:300 for immunofluorescence in various studies.

Biochem/physiol Actions

IgG1 class is the most abundant and its deficiency results in hypogammaglobulinemia. IgG2 deficiency increases susceptibility to bacterial infections. IgG3 mediates effector functions. IgG4 is associated with asymptomatic infection. Digestion of IgG by papain results in generation of fragment antigen binding (Fab). Pepsin digestion of IgG results in fragment crystallizable (Fc). The Fc region of IgG antibody has enormous therapeutic potential and is exploited for the development of therapeutic antibodies.
Specificity for the Fab fragment of mouse IgG is determined by immunoelectrophoresis (IEP). The conjugate shows no reactivity with mouse IgG, Fc fragment, or human IgG.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Preparation Note

Adsorbed to reduce background staining with human samples.
For continuous use, store at 2-8 °C for a maximum of one month. For extended storage, the solution may be frozen in working aliquots. Repeated freezing and thawing, or storage in "frost-free" freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

or

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content

Instructions


Mechanism of action of cytotoxic compounds from the seeds of Euphorbia lathyris.
Teng YN, et al.
Phytomedicine, 41(3), 62-66 (2018)
Sitagliptin and Roux-en-Y gastric bypass modulate insulin secretion via regulation of intra-islet PYY.
Guida C, et al.
Diabetes, obesity & metabolism, 20(3), 571-581 (2018)
Khaled Khoufache et al.
Medical mycology, 48(8), 1049-1055 (2010-07-09)
Since the airway epithelium is the first tissue encountered by airborne fungal spores, specific models are needed to study this interaction. We developed such a model using primary porcine tracheal epithelial cells (PTEC) as a possible alternative to the use



Global Trade Item Number

SKUGTIN
F5262-.5ML04061833618158
F5262-1ML04061833618165