Skip to Content
Merck
All Photos(1)

Key Documents

F5883

Sigma-Aldrich

4-Fluoro-7-nitrobenzofurazan

≥98% (elemental analysis)

Synonym(s):

NBD-F

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H2FN3O3
CAS Number:
Molecular Weight:
183.10
Beilstein:
1077571
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.32

Quality Level

Assay

≥98% (elemental analysis)

mp

52-54 °C (lit.)

fluorescence

λex 380 nm; λem 515 nm (hydrolyzed (esterase))

storage temp.

2-8°C

SMILES string

[O-][N+](=O)c1ccc(F)c2nonc12

InChI

1S/C6H2FN3O3/c7-3-1-2-4(10(11)12)6-5(3)8-13-9-6/h1-2H

InChI key

PGZIDERTDJHJFY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Useful for fluorescent labeling of amino acids and amines in HPLC

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Yang-Ki Park et al.
Journal of chromatography. A, 1217(20), 3357-3361 (2010-04-13)
Single drop microextraction (SDME) can be in-line coupled with capillary electrophoresis by attaching a drop to the tip of a capillary. With a 2-layer drop comprised of an aqueous basic acceptor phase covered with a thin organic layer, acidic analytes
Yanting Song et al.
Amino acids, 42(5), 1897-1902 (2011-04-21)
A high-performance liquid chromatography method in which fluorescence detection is used for the simultaneous determination of 21 amino acids is proposed. Amino acids were derivatized with 4-fluoro-7-nitro-2,1,3-benzoxadiazole (NBD-F) and then separated on a monolithic silica column (MonoClad C18-HS, 150 mm×3
Hua-Ming Tseng et al.
Electrophoresis, 30(8), 1399-1405 (2009-03-26)
A sensitive CE with LIF method has been developed for quantitative analysis of small carbohydrates. In this work, 17 carbohydrates including mono-, di- and oligosaccharides were simultaneously derivatized with 4-fluoro-7-nitrobenzofurazane (NBD-F) via a two-step reaction involving reductive amination with ammonia
Chiaki Aoyama et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 25(1), 63-65 (2009-01-14)
A simple and sensitive determination method for selenomethionine (Se-Met) using an HPLC-fluorescence detection system coupled with an on-line electrochemical reactor has been developed. NBD-F (4-fluoro-7-nitro-2,1,3-benzoxadiazole) was used as the fluorescent derivatization reagent for Se-Met. NBD-Se-Met was separated from NBD-derivatives of
Nasr Y Khalil
Talanta, 80(3), 1251-1256 (2009-12-17)
A fully automated, rapid and highly sensitive HPLC method with automated sample pre-treatment by column-switching system and fluorescence detection has been developed for the trace quantitative determination of the new antidepressant reboxetine (RBX) in human plasma. A simple pre-column derivatization

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service