Skip to Content
Merck
All Photos(2)

Documents

471305

Sigma-Aldrich

Butylamine

99.5%

Synonym(s):

1-Aminobutane, n-Butylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3NH2
CAS Number:
Molecular Weight:
73.14
Beilstein:
605269
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

vapor density

2.5 (vs air)

Quality Level

vapor pressure

68 mmHg ( 20 °C)

Assay

99.5%

autoignition temp.

594 °F

expl. lim.

9.8 %

refractive index

n20/D 1.401 (lit.)

bp

78 °C (lit.)

mp

−49 °C (lit.)

solubility

water: miscible

density

0.74 g/mL at 25 °C (lit.)

SMILES string

CCCCN

InChI

1S/C4H11N/c1-2-3-4-5/h2-5H2,1H3

InChI key

HQABUPZFAYXKJW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Butylamine (n-Butylamine, BuA) is a primary aliphatic amine. Its pyrolysis mechanism has been investigated. BuA can be synthesized from butyronitrile via Co/SiO2 (silicon dioxide) assisted hydrogenation. A decrease in fluorescence intensity was observed on adding BuA to a solution of colloidal CdSe nanoparticles (NPs).

Application

Butylamine (n-Butylamine) may be used as a template to generate hierarchical monolithic zeolites. It may also be used as a dispersion medium for fluorinated graphene sheets (F-GSs) to generate butylamine modified F-GSs.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Met. Corr. 1 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

19.4 °F - closed cup

Flash Point(C)

-7 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation of ZSM-5 Coated on Monolithic Interconnected Macroporous Al2O3 Using Cheap n-Butylamine as Template.
Sun M, et al.
Chin. J. Chem., 33(9), 1057-1063 (2015)
Impact of solvent on Co/SiO2 activity and selectivity for the synthesis of n-butylamine from butyronitrile hydrogenation.
Segobia DJ, et al.
Catalysis Communications, 62, 62-66 (2015)
Hydrogenation of nitriles to primary amines on metal-supported catalysts: Highly selective conversion of butyronitrile to n-butylamine.
Segobia DJ, et al.
Applied Catalysis A: General, 445, 69-75 (2012)
The Thermal Decomposition of n-Butylamine.
Beachell HC and Taylor HA.
J. Chem. Phys. , 10(2), 106-110 (1942)
Use of butylamine modified graphene sheets in polymer solar cells.
Valentini, L, et al.
Journal of Materials Chemistry, 20(5), 995-1000 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service