Skip to Content
Merck
All Photos(3)

Key Documents

F3639

Sigma-Aldrich

4-Fluoro-7-sulfamoylbenzofurazan

Synonym(s):

4-(Aminosulfonyl)-7-fluorobenzofurazane, 7-Fluoro-2,1,3-benzoxadiazole-4-sulfonamide, 7-Fluorobenzofurazan-4-sulfonamide, ABD-F

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4FN3O3S
CAS Number:
Molecular Weight:
217.18
Beilstein:
5035147
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.32

storage temp.

−20°C

Quality Level

SMILES string

NS(=O)(=O)c1ccc(F)c2nonc12

InChI

1S/C6H4FN3O3S/c7-3-1-2-4(14(8,11)12)6-5(3)9-13-10-6/h1-2H,(H2,8,11,12)

InChI key

XROXHZMRDABMHS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Fluoro-7-sulfamoylbenzofurazan is a fluorescent reagent, which directly links to the sulfur atom of thiols without a flexible alkyl chain. It is used to detect low molecular thiols using high performance liquid chromatography.

Application

Reagent for fluorimetric assay of thiols.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

C C Chin et al.
Analytical biochemistry, 214(1), 128-134 (1993-10-01)
The use of the reagent tributyl phosphine (Bu3P) to reduce disulfides (Ruegg, U.T., and Rudinger, J., Methods Enzymol. 47, 111-116, 1977) and of 4-(aminosulfonyl)-7-fluoro-2,1,3-benzoxadiazole (ABD-F) to block free sulphydryl groups (Toyo'oka, T., and Imai, K. Anal. Chem. 56, 2461-2464, 1984)
Chie Moritani et al.
Biological & pharmaceutical bulletin, 40(7), 984-991 (2017-04-07)
Glutathione (GSH) is an ubiquitous thiol-containing tripeptide, which plays important roles in cellular protection from oxidative stress. In our search for a dietary source that can increase GSH levels, we discovered that a 24 h treatment of HepG2 cells with rice
T Hiratsuka
The Journal of biological chemistry, 268(33), 24742-24750 (1993-11-25)
4-Fluoro-7-sulfamoylbenzofurazan (ABDF) has a rather small fluorophore that is linked directly to the sulfur atom of thiols without a flexible alkyl chain (Imai, K., and Toyo'oka, T. (1987) Methods Enzymol. 143, 67-75). In the present study I examined the fluorescent
Mikhail Linetsky et al.
Investigative ophthalmology & visual science, 44(9), 3920-3926 (2003-08-27)
To determine the mechanism that leads to the UVA light-dependent loss of glutathione reductase (GR) activity in human lens (HL). Both the HL water-soluble (WS) fraction and yeast GR were irradiated with UVA light (200 mW/(cm(2). h) for 1 hour
James E Noble et al.
Analytical chemistry, 75(9), 2042-2047 (2003-05-02)
A homogeneous microplate assay for the serine/threonine protein phosphatases PP1 and PP2A, employing fluorescent-labeled phosphopeptides, has been developed. Phosphopeptides derived from a phosphoacceptor site in myelin basic protein were designed with a cysteine adjacent to the phosphoresidue, allowing site-selective labeling

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service