Skip to Content
Merck
All Photos(3)

Documents

F2555

Sigma-Aldrich

Monoclonal ANTI-FLAG® antibody produced in rabbit

clone SIG1-25, ascites fluid

Synonym(s):

Anti-ddddk, Anti-dykddddk

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.32

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

SIG1-25, monoclonal

technique(s)

immunocytochemistry: 1:125-1:250 using transiently transfected cells expressing FLAG (sequence at the N-terminus)-tagged protein fixed with paraformaldehyde/Triton X-100
indirect ELISA: suitable
western blot: 1:250-1:500 using extracts of transiently transfected cells expressing FLAG (sequence at the N-terminus)-tagged protein

isotype

IgG

immunogen sequence

DYKDDDDK

shipped in

dry ice

storage temp.

−20°C

General description

Epitope tags provide a method to localize gene products in a variety of cell types, study the topology of proteins and protein complexes, identify associated proteins, and characterize newly identified, low abundance, or poorly immunogenic proteins when protein specific antibodies are not available. Tagging with the FLAG® peptide sequence may be done at the N-terminus, N-terminus preceded by a methionine residue, C-terminus, or at internal positions of the target protein. FLAG may also be placed in association with other tags. The small size of the FLAG tag or sequence and its high hydrophilicity tend to decrease the possibility of interference with the protein expression, proteolytic maturation, antigenicity, and function. The N-terminal FLAG® peptide sequence contains a unique enterokinase cleavage site allowing it to be completely removed from the purified fusion proteins. Cleavage catalyzed by Cu2+ ions of the C-terminal FLAG® peptide from a fusion protein has been reported. A sequence motif with five out of eight amino acid residues identical to the FLAG® peptide is found in both rat and mouse Mg2+ dependent protein b-phosphatase, as well as in the human and bovine enzyme. This rabbit ANTI-FLAG® antibody is produced from the hybridoma SIG1-25. The hybridoma is derived by the fusion of myeloma cells and splenocytes from rabbits immunized with the FLAG® peptide sequence conjugated to KLH.

Specificity

This monoclonal antibody recognizes the FLAG sequence at the N-terminus

Immunogen

FLAG peptide sequence DYKDDDDK

Application

Monoclonal ANTI-FLAG® antibody produced in rabbit has been used in:

  • immunofluorescence
  • western blot analysis
  • immunocytochemistry
  • indirect ELISA

Learn more product details in our FLAG® application portal.

Physical form

Provided as ascites fluid containing 15 mM sodium azide as a preservative.

Legal Information

ANTI-FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany
FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany
Triton is a trademark of The Dow Chemical Company or an affiliated company of Dow

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Recombinant lentivirus with enhanced expression of caudal-related homeobox protein 2 inhibits human colorectal cancer cell proliferation in vitro
He S, et al.
Molecular Medicine Reports, 12(2), 1838-1844 (2015)
Qungang Qi et al.
The Journal of biological chemistry, 287(37), 31482-31493 (2012-07-18)
Previous studies have demonstrated that Arabidopsis thaliana BBX32 (AtBBX32) represses light signaling in A. thaliana and that expression of AtBBX32 in soybean increases grain yield in multiple locations and multiyear field trials. The BBX32 protein is a member of the
Yury A Bochkov et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(17), 5485-5490 (2015-04-08)
Members of rhinovirus C (RV-C) species are more likely to cause wheezing illnesses and asthma exacerbations compared with other rhinoviruses. The cellular receptor for these viruses was heretofore unknown. We report here that expression of human cadherin-related family member 3
Increased protein stability of CDKN1C causes a gain-of-function phenotype in patients with IMAGe syndrome
Hamajima N, et al.
PLoS ONE, 8(9), e75137-e75137 (2013)
Reduction of NANOG mediates the inhibitory effect of aspirin on tumor growth and stemness in colorectal cancer
Wang H, et al.
Cellular Physiology and Biochemistry, 44(3), 1051-1063 (2017)

Related Content

Protein and nucleic acid interaction reagents and resources for investing protein-RNA, protein-DNA, and protein-protein interactions and associated applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service