ET0200
E-TOXATE™ Kit
sufficient for 20 assays
Synonym(s):
Endotoxin detection kit
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352202
NACRES:
NA.84
Recommended Products
usage
sufficient for 20 assays
storage temp.
2-8°C
General description
E-Toxate™ Kit prepared from Limulus polyphemus amebocyte lysate (LAL) is suitable for the detection and quantification of endotoxins for research purposes.
Application
E-Toxate™ Kit has been used to determine the endotoxins:
- from Tabebuia chrysantha and Tabebuia rosea plant methanolic extracts
- from human pentameric C-reactive protein (pCRP)
- in the Diptheria toxin preparations
The E-TOXATE Kit was used to determine the number of bacteria in suspensions of group B streptococci used to infect rats for models of sepsis.
Features and Benefits
- Highly sensitive to endotoxins
- Simple and rapid detection
- Cost-effective
Legal Information
E-Toxate is a trademark of Sigma-Aldrich Co. LLC
comparable product
Product No.
Description
Pricing
related product
Product No.
Description
Pricing
Storage Class Code
12 - Non Combustible Liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Priscila Raquel Martins et al.
Memorias do Instituto Oswaldo Cruz, 103(3), 244-250 (2008-06-12)
A polysaccharide-rich fraction (ATF) of medicinal mushroom Agaricus brasiliensis was evaluated on the candidacidal activity, H2O2 and nitric oxide (NO) production, and expression of mannose receptors by murine peritoneal macrophages. Mice received three intraperitoneal (i.p.) injections of ATF and after
Priscila Raquel Martins et al.
The journal of venomous animals and toxins including tropical diseases, 23, 17-17 (2017-03-28)
Agaricus brasiliensis is a medicinal mushroom with immunomodulatory and antitumor activities attributed to the β-glucans presented in the polysaccharide fraction of its fruiting body. Since β-glucans enhance cellular immunoresponsiveness, in this study we aimed to evaluate the effect of an
Anne Kiang et al.
Molecular therapy : the journal of the American Society of Gene Therapy, 14(4), 588-598 (2006-05-31)
Excessive complement activation can result in extreme tissue damage and systemic inflammatory responses, similar to innate immune responses rapidly elicited after systemic adenovirus (Ad) injections. To determine if Ad interactions with the complement system impact upon Ad-induced innate immune responses
Jami E Milam et al.
The American journal of pathology, 176(1), 218-226 (2009-12-02)
To investigate the role of CD11c(+) cells in endotoxin-induced acute lung injury, wild-type or CD11c-diphtheria toxin receptor transgenic mice were treated with intraperitoneal diphtheria toxin (5 ng/g b.wt.) in the presence or absence of intratracheal lipopolysaccharide (51 microg). Lipopolysaccharide treatment
Jean-Pierre Louboutin et al.
Antioxidants (Basel, Switzerland), 3(2), 414-438 (2014-01-01)
HIV-1 trans-acting protein Tat, an essential protein for viral replication, is a key mediator of neurotoxicity. If Tat oxidant injury and neurotoxicity have been described, consequent neuroinflammation is less understood. Rat caudate-putamens (CPs) were challenged with Tat, with or without
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service