Skip to Content
Merck

Skip To

ET0100

E-Toxate Kit

sufficient for 50 assays

Synonym(s):

Endotoxin detection kit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.83
UNSPSC Code:
12352202
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


usage

sufficient for 50 assays

Quality Segment

storage temp.

2-8°C

General description

E-Toxate Kit made from Limulus polyphemus amebocyte lysate (LAL) is developed for the detection and semi-quantitation of endotoxins.

Application

E-TOXATE Kit was used to determine the number of bacteria in suspensions of group B streptococci used to infect rats for models of sepsis.
E-Toxate Kit has been used for endotoxin detection:
  • in chitosan and collagen solutions
  • in C-reactive protein (CRP) samples
  • in the ethanolic plant extracts from Tinospora cordifolia

Features and Benefits

  • Highly sensitive to endotoxins
  • Simple and rapid detection
  • Cost-effective

Legal Information

E-Toxate is a trademark of Sigma-Aldrich Co. LLC

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
ET0200PLED35NA0410
description

sufficient for 50 assays

description

sufficient for 20 assays

description

sufficient for 35 purifications

description

sufficient for 25 preparations

usage

sufficient for 50 assays

usage

sufficient for 20 assays

usage

sufficient for 35 purifications

usage

sufficient for 25 preparations

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

15-25°C

storage temp.

15-25°C


Still not finding the right product?

Explore all of our products under E-Toxate Kit


Kit Components Also Available Separately

Product No.
Description
SDS & Pricing

  • E-Toxate Endotoxin standard, for endotoxin quantitation 1 mL/vial
    SDS

  • E-Toxate reagent from Limulus polyphemus, sufficient for 50 tests 1 mL/vial
    SDS

  • E-Toxate Water, endotoxin, free 30 mL
    SDS

wgk

WGK 3

Storage Class

12 - Non Combustible Liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters