Skip to Content
Merck
All Photos(1)

Documents

E5630

Sigma-Aldrich

β-Estradiol 6-(O-carboxy­methyl)oxime: BSA

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

sterility

non-sterile

Quality Level

form

lyophilized powder

extent of labeling

~30 mol steroid per mol BSA

solubility

phosphate buffer: 0.90-1.10 mg/mL, faintly hazy to hazy, colorless to faintly yellow

shipped in

ambient

storage temp.

2-8°C

General description

β-Estradiol 6-(O-carboxy­methyl)oxime: BSA is an estradiol conjugate and is membrane-impermeable. BSA is linked to the estradiol at position 6 to form the β-estradiol 6-(O-carboxymethyl) oxime: BSA conjugate.

Application

β-Estradiol 6-(O-carboxy­methyl)oxime: BSA has been used:
  • to test its effect on the N-methyl-D-aspartate receptor (NMDAR) currents in female dorsal root ganglion (DRG) neurons
  • to coat microplates for the detection of plasma 17β-Estradiol levels using indirect enzyme-linked immunosorbent assay (ELISA)
  • to test its effect on MutL homolog 1 in colorectal cancer lines

Biochem/physiol Actions

β-Estradiol 6-(O-carboxy­methyl)oxime: BSA mimics estrogen and favors amyloid precursor protein α (sAPPα) secretion in vitro studies with primary rat cortical neurons. It also stimulates mitogen-activated protein kinase (MAPK) activity in human umbilical vein endothelial cells

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sex-dependent differences in the activity and modulation of N-methyl-d-aspartic acid receptors in rat dorsal root ganglia neurons.
McRoberts, et al.
Neuroscience, 148, 1015-1020 (2018)
Adrián Tintos et al.
Ecotoxicology and environmental safety, 66(2), 139-147 (2006-02-10)
To assess the effects of naphthalene on liver intermediary metabolism and plasma steroid hormones, immature female rainbow trout (Oncorhynchus mykiss), in a first experiment, were intraperitoneally injected (2 microLg(-1)) with vegetable oil alone (control) or containing naphthalene (10 and 50
Yu-Jie Lai et al.
Journal of molecular neuroscience : MN, 61(3), 436-448 (2017-02-06)
Estrogen exerts multiple actions in the brain and is an important neuroprotective factor in a number of neuronal disorders. However, the underlying mechanism remains unknown. Studies demonstrate that ubiquitin-conjugating enzyme 9 (ubc9) has an integral role in synaptic plasticity and
Sun Zhang et al.
Acta pharmacologica Sinica, 26(2), 171-176 (2005-01-25)
To investigate the mechanism of the action of estrogen, which stimulates the release of secreted amyloid precursor protein alpha (sAPP(alpha)) and decreases the generation of amyloid-beta protein (A(beta)), a dominant component in senile plaques in the brains of Alzheimer's disease
Claudio R F Marinho et al.
PloS one, 4(5), e5630-e5630 (2009-05-23)
Pregnancy-associated malaria (PAM) is associated with placenta pathology and poor pregnancy outcome but the mechanisms that control the malaria parasite expansion in pregnancy are still poorly understood and not amenable for study in human subjects. Here, we used a set

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service