Skip to Content
Merck
All Photos(1)

Key Documents

E3263

Sigma-Aldrich

Efaroxan hydrochloride

≥98% (HPLC)

Synonym(s):

2-Ethyl-2-(imidazolin-2-yl)-2,3-dihydrobenzofuran hydrochloride, RX 821037A

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H16N2O · HCl
CAS Number:
Molecular Weight:
252.74
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

solubility

H2O: >20 mg/mL

SMILES string

Cl.CCC1(Cc2ccccc2O1)C3=NCCN3

InChI

1S/C13H16N2O.ClH/c1-2-13(12-14-7-8-15-12)9-10-5-3-4-6-11(10)16-13;/h3-6H,2,7-9H2,1H3,(H,14,15);1H

InChI key

DWOIUCRHVWIHAH-UHFFFAOYSA-N

Related Categories

Biochem/physiol Actions

(+)-Isomer is a selective α-adrenoceptor antagonist; (−)-isomer is an imidazoline ligand that induces insulin secretion, mediated by the blockade of ATP-sensitive potassium channels in pancreatic β cells; I1 imidazoline binding site antagonist.
(+)-Isomer is a selective α-adrenoceptor antagonist; (−)-isomer is an imidazoline ligand; I1 imidazoline binding site antagonist.

Features and Benefits

This compound is featured on the Imidazoline Binding Sites and Potassium Channels pages of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I I Zaitseva et al.
Cellular and molecular life sciences : CMLS, 65(7-8), 1248-1255 (2008-02-19)
An insufficient number of insulin-producing beta-cells is a major cause of defective control of blood glucose in both type 1 and type 2 diabetes. The aim of this study was to clarify whether the insulinotropic imidazolines can affect the survival
Shaifali Bhalla et al.
Pharmacology, biochemistry, and behavior, 103(3), 550-560 (2012-10-30)
The potentiation of oxycodone antinociception by BMS182874 (endothelin-A (ET(A)) receptor antagonist) and agmatine (imidazoline receptor/α(2)-adrenoceptor agonist) is well-documented. It is also known that imidazoline receptors but not α(2)-adrenoceptors are involved in potentiation of oxycodone antinociception by agmatine and BMS182874 in
J C Chapman et al.
Diabetes, 48(12), 2349-2357 (1999-12-02)
The properties of ATP-sensitive K+ (K(ATP)) channels were explored in the electrofusion-derived, glucose-responsive, insulin-secreting cell line BRIN-BD11 using patch-clamp techniques. In intact cells, K(ATP) channels were inhibited by glucose, the sulfonylurea tolbutamide, and the imidazoline compounds efaroxan and phentolamine. Each
Fei Li et al.
Addiction biology, 17(2), 392-408 (2011-10-05)
Agmatine, an endogenous ligand for imidazoline receptor, has been shown to prevent opioid dependence, but not much is known about the mechanisms of the effect of agmatine. In the present study, we investigated the function of I1 imidazoline receptor and
M A Carlisle et al.
The Journal of pharmacology and experimental therapeutics, 274(2), 598-601 (1995-08-01)
I1-imidazoline receptor activation by moxonidine has potent antigastric secretory and gastroprotective effects in rats. We therefore tested whether an imidazoline receptor antagonist, efaroxan, would influence gastric secretion and block the antisecretory and antiulcer effects of moxonidine. When given intracerebroventricularly (i.c.v.)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service