Skip to Content
Merck
All Photos(1)

Key Documents

A6566

Sigma-Aldrich

2-Hydroxysaclofen

≥98% (TLC), solid

Synonym(s):

3-Amino-2-(4-chlorophenyl)-2-hydroxypropanesulfonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H12ClNO4S
CAS Number:
Molecular Weight:
265.71
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32

Quality Level

Assay

≥98% (TLC)

form

solid

color

white

solubility

H2O: 1 mg/mL
0.1 M HCl: 1.2 mg/mL
methanol: 1.4 mg/mL
DMSO: 28 mg/mL
0.1 M NaOH: 9.5 mg/mL

SMILES string

NCC(O)(CS(O)(=O)=O)c1ccc(Cl)cc1

InChI

1S/C9H12ClNO4S/c10-8-3-1-7(2-4-8)9(12,5-11)6-16(13,14)15/h1-4,12H,5-6,11H2,(H,13,14,15)

InChI key

WBSMZVIMANOCNX-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Potent and selective antagonist at GABAB receptors.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D R Curtis et al.
Neuroscience letters, 92(1), 97-101 (1988-09-23)
When administered microelectrophoretically, a sulphonic acid derivative of baclofen, 3-amino-2-(4-chlorophenyl)-2-hydroxy-propylsulphonic acid, reversibly reduced the presynaptic reduction by (-)-baclofen of the monosynaptic excitation of spinal interneurones by impulses in low threshold primary afferent fibres of the cat as well as the
K Obrietan et al.
Journal of neurophysiology, 82(1), 94-102 (1999-07-13)
In the mature nervous system excitatory neurotransmission mediated by glutamate is balanced by the inhibitory actions of GABA. However, during early development, GABA acting at the ligand-gated GABAA Cl- channel also exerts excitatory actions. This raises a question as to
Jacques H Abraini et al.
Anesthesia and analgesia, 96(3), 746-749 (2003-02-25)
Inhaled anesthetics, including the gaseous anesthetics nitrous oxide and xenon, are thought to act by interacting directly with ion-channel receptors. In contrast, little is known about the mechanism of action of inert gases that show only narcotic potency at high
Nonna A Otmakhova et al.
Journal of neurophysiology, 92(4), 2027-2039 (2004-05-28)
CA1 pyramidal cells receive two major excitatory inputs: the perforant path (PP) terminates in the most distal dendrites, whereas the Schaffer collaterals (SC) terminate more proximally. We have examined the mechanism of the afterhyperpolarization (AHP) that follows single subthreshold excitatory
K Seki et al.
The Journal of physiology, 533(Pt 2), 503-518 (2001-06-05)
1. To investigate the mechanisms for the coding stimulus sequence in the auditory cortex (AC), post-tetanic potentiation (PTP) was recorded after sequentially combined heterosynaptic stimulation was applied in rat AC slices. 2. Brief tetanic stimulation (TS) was applied at two

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service