Skip to Content
Merck
All Photos(1)

Key Documents

SML0591

Sigma-Aldrich

Chlorthalidone

≥98% (HPLC)

Synonym(s):

2-Chloro-5(1-hydroxy-3-oxo-1- isoindolinyl) benzenesulfonamide, 2-Chloro-5-(2,3-dihydro-1-hydroxy-3-oxo-1H-isoindol-1-yl)benzenesulfonamide, Chlortalidone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H11ClN2O4S
CAS Number:
Molecular Weight:
338.77
EC Number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

[S](=O)(=O)(N)c1c(ccc(c1)C2(NC(=O)c3c2cccc3)O)Cl

InChI

1S/C14H11ClN2O4S/c15-11-6-5-8(7-12(11)22(16,20)21)14(19)10-4-2-1-3-9(10)13(18)17-14/h1-7,19H,(H,17,18)(H2,16,20,21)

InChI key

JIVPVXMEBJLZRO-UHFFFAOYSA-N

Biochem/physiol Actions

Chlorthalidone is a thiazide-like diuretic, an inhibitor of the Na+-Cl- cotransporter.
Chlorthalidone is a thiazide-like diuretic, an inhibitor of the Na+-Cl- cotransporter. Chlorthalidone inhibits sodium ion transport across the renal tubular epithelium increasing the delivery of sodium to the distal renal tubule and indirectly increasing potassium excretion via the sodium-potassium exchange mechanism. Chlorthalidone also promotes Ca++ reabsorption by an unknown mechanism. Several recent comparison studies inidcate that chlorthalidone may be a better drug in preventing cardiovascular events than hydrochlorothiazide.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Feras Karadsheh et al.
Current hypertension reports, 14(5), 416-420 (2012-08-14)
Thiazide and thiazide-like diuretics have been widely used as blood pressure-lowering agents for more than 5 decades. However, their use in patients with advanced chronic kidney disease has been limited and often discouraged. The exact mechanism of how thiazide and
William C Cushman et al.
Hypertension (Dallas, Tex. : 1979), 60(2), 310-318 (2012-06-20)
Azilsartan medoxomil, an effective, long-acting angiotensin II receptor blocker, is a new treatment for hypertension that is also being developed in fixed-dose combinations with chlorthalidone, a potent, long-acting thiazide-like diuretic. We compared once-daily fixed-dose combinations of azilsartan medoxomil/chlorthalidone force titrated
Michael Proschan et al.
Statistics in medicine, 32(5), 884-897 (2012-09-11)
The debate over whether certain antihypertensive medications have benefits beyond what would be expected from their blood pressure lowering spurred the Antihypertensive and Lipid-Lowering Treatment to Prevent Heart Attack Trial, which randomized 42,418 participants to chlorthalidone (15,255), amlodipine (9048), lisinopril
Brian C Lund et al.
Journal of clinical hypertension (Greenwich, Conn.), 14(9), 623-629 (2012-09-06)
Indirect evidence suggests that chlorthalidone may be more effective than hydrochlorothiazide (HCTZ), but direct comparisons are lacking. Using national Veterans Administrative pharmacy data from 2003 to 2008, the authors performed a retrospective cohort study examining the effectiveness of chlorthalidone and
Domenic Sica et al.
Journal of clinical hypertension (Greenwich, Conn.), 14(5), 284-292 (2012-04-27)
This study compared the efficacy and safety of fixed-dose combinations (FDCs) of the angiotensin II receptor blocker azilsartan medoxomil (AZL-M) and the thiazide-like diuretic chlorthalidone (CLD) with the individual monotherapies in a double-blind factorial study. A total of 1714 patients

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service