Skip to Content
Merck
All Photos(2)

Key Documents

E0516

Sigma-Aldrich

Etodolac

Synonym(s):

1,8-Diethyl-1,3,4,9-tetrahydropyrano[3,4-b]indole-1-acetic acid, Etodolic acid, NSC 282126

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H21NO3
CAS Number:
Molecular Weight:
287.35
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

originator

Wyeth

storage temp.

2-8°C

SMILES string

CCc1cccc2c3CCOC(CC)(CC(O)=O)c3[nH]c12

InChI

1S/C17H21NO3/c1-3-11-6-5-7-12-13-8-9-21-17(4-2,10-14(19)20)16(13)18-15(11)12/h5-7,18H,3-4,8-10H2,1-2H3,(H,19,20)

InChI key

NNYBQONXHNTVIJ-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Non-steroidal anti-inflammatory compound that is a non-selective inhibitor of COX-1 and COX-2.

Features and Benefits

This compound was developed by Wyeth. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Zvaifler
Clinical rheumatology, 8 Suppl 1, 43-53 (1989-03-01)
Etodolac (Lodine, Ramodar, Ultradol), an anti-inflammatory, analgesic agent, is the first of a new class of nonsteroidal anti-inflammatory drugs (NSAIDs), the pyranocarboxylic acids. A review of the literature on numerous clinical studies showed that etodolac (200 to 600 mg/day) is
Yael Goldfarb et al.
Annals of surgery, 253(4), 798-810 (2011-04-09)
Surgical procedures, including primary tumor resection, have been suggested to suppress immune competence and to promote postoperative infections and cancer metastasis. Catecholamines and prostaglandins were recently implicated in these processes, and in directly promoting tumor angiogenesis and invasion. To examine
S Lynch et al.
Drugs, 31(4), 288-300 (1986-04-01)
Etodolac is a new non-steroidal agent (NSAID) with anti-inflammatory and analgesic activity. It has been studied in rheumatoid arthritis, osteoarthritis, and pain resulting from minor surgery. In the latter setting, etodolac is at least as effective as aspirin in terms
D C Brater et al.
Clinical rheumatology, 8 Suppl 1, 25-35 (1989-03-01)
The pharmacokinetics of etodolac, a new nonsteroidal anti-inflammatory drug, were compared in normal subjects, in patients with renal and hepatic disease, and in elderly patients. In 28 normal subjects, orally administered etodolac was rapidly absorbed. By 1.2 hours after ingestion
Shravan Kumar Tirunagari et al.
The Cochrane database of systematic reviews, (3)(3), CD007357-CD007357 (2009-07-10)
Etodolac is a selective cyclo-oxygenase-2 (COX-2) inhibitor, with evidence of efficacy in osteoarthritis and rheumatoid arthritis. Its analgesic efficacy in postoperative pain has not been clearly established. There are no systematic reviews on Etodolac's use in this condition. To assess

Articles

Discover Bioactive Small Molecules for Lipid Signaling Research

Discover Bioactive Small Molecules for Lipid Signaling Research

Discover Bioactive Small Molecules for Lipid Signaling Research

Discover Bioactive Small Molecules for Lipid Signaling Research

Chromatograms

application for HPLC

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service