Skip to Content
Merck
All Photos(1)

Key Documents

K1751

Sigma-Aldrich

Ketoprofen

≥98% (TLC), powder, non-steroidal anti-inflammatory compound

Synonym(s):

2-(3-Benzoylphenyl)propionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H14O3
CAS Number:
Molecular Weight:
254.28
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Product Name

Ketoprofen, ≥98% (TLC)

biological source

synthetic

Assay

≥98% (TLC)

form

powder

solubility

ethanol: 50 mg/mL, clear, colorless to yellow

originator

Bayer

SMILES string

CC(C(O)=O)c1cccc(c1)C(=O)c2ccccc2

InChI

1S/C16H14O3/c1-11(16(18)19)13-8-5-9-14(10-13)15(17)12-6-3-2-4-7-12/h2-11H,1H3,(H,18,19)

InChI key

DKYWVDODHFEZIM-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Ketoprofen belongs to the class of 2-arylpropionic acid.

Application

Ketoprofen has been used:
  • as sample to analyse the effects of long storage period by chromatographic and microscopic techniques
  • as a nonselective COX inhibitor to inject subcutaneously in rats to study its effect on stress/methamphetamine hydrochloride-induced changes in occludin, claudin-5 and COX-2 protein immunoreactivity, truncation of β-dystroglycan, brain water content and fluorescein isothiocyanate-dextran extravasation
  • in phosphate buffer to study its ability to inhibit heat-induced denaturation of albumin

Biochem/physiol Actions

It serves as an efficient drug to treat ankylosing spondylitis, rheumatoid arthritis and osteoarthritis. It also has antipyretic and analgesic effects. Ketoprofen prevents the action of prostaglandin synthetase.
Non-steroidal anti-inflammatory compound that is selective for COX-1.

Features and Benefits

This compound was developed by Bayer. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Polyamidoamine dendrimers used as solubility enhancers of ketoprofen
Yiyun C, et al.
European Journal of Medicinal Chemistry, 40(12), 1390-1393 (2005)
Ekaterina Blinova et al.
Biomolecules, 11(6) (2021-07-03)
The main goal of our study was to explore the wound-healing property of a novel cerium-containing N-acethyl-6-aminohexanoate acid compound and determine key molecular targets of the compound mode of action in diabetic animals. Cerium N-acetyl-6-aminohexanoate (laboratory name LHT-8-17) as a
Ekaterina Blinova et al.
International journal of molecular sciences, 22(18) (2021-09-29)
The possible involvement of p53 signaling, FGFR3 expression, and FGFR3 mutation rates in the prediction of the NMIBC anti-PD-L1 treatment response needs to be clarified. The main aim of our study was to explore predictive value of p53 expression, FGFR3
In vitro chiral conversion, phase separation, and wave propagation in aged profen solutions
Sajewicz M, et al.
Journal of Liquid Chromatography and Related Technologies, 32(9), 1359-1372 (2009)
L Saso et al.
Archives of pharmacal research, 24(2), 150-158 (2001-05-08)
The activity of nonsteroidal antiinflammatory drugs (NSAIDs) in rheumatoid arthritis is not only due to the inhibition of the production of prostaglandins, which can even have beneficial immunosuppressive effects in chronic inflammatory processes. Since we speculated that these drugs could

Articles

Human epithelial intestinal colonic organoids can be used as an alternative to Caco-2 drug permeability assays for drug screening and compound toxicity testing.

Discover Bioactive Small Molecules for Lipid Signaling Research

Discover Bioactive Small Molecules for Lipid Signaling Research

Discover Bioactive Small Molecules for Lipid Signaling Research

See All

Chromatograms

application for HPLCapplication for HPLCapplication for HPLC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service