Skip to Content
Merck
All Photos(1)

Key Documents

M3935

Sigma-Aldrich

Meloxicam sodium salt hydrate

≥98% (HPLC)

Synonym(s):

4-Hydroxy-2-methyl-N-(5-methyl-2-thiazolyl)-2H-1,2-benzothiazine-3-carboxamide 1,1-dioxide sodium hydrate, Metacam sodium salt hydrate, Mobec sodium salt hydrate, UH-AC 62XX sodium salt hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H12N3NaO4S2 · xH2O
CAS Number:
Molecular Weight:
373.38 (anhydrous basis)
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Assay

≥98% (HPLC)

form

powder

color

light yellow to dark yellow

solubility

DMSO: 5 mg/mL, clear

originator

Boehringer Ingelheim

storage temp.

room temp

SMILES string

O.[Na+].CN1C(C(=O)Nc2ncc(C)s2)=C([O-])c3ccccc3S1(=O)=O

InChI

1S/C14H13N3O4S2.Na.H2O/c1-8-7-15-14(22-8)16-13(19)11-12(18)9-5-3-4-6-10(9)23(20,21)17(11)2;;/h3-7,18H,1-2H3,(H,15,16,19);;1H2/q;+1;/p-1

InChI key

IZBZAOZGNNQKPJ-UHFFFAOYSA-M

Gene Information

human ... PTGS2(5743)

Application

Meloxicam sodium salt hydrate has been used:
  • as a cyclooxygenase-2 (COX-2) inhibitor in solid Ehrlich tumor[1]
  • as an alternative to diclofenac nonsteroidal anti-inflammatory drugs (NSAID) to treat vultures[2]
  • as a reference standard in liquid chromatography electrospray ionisation mass spectrometry (LC-ESI/MS)[3]
  • to test its effect on prostaglandin (PGE2) production in lipopolysaccharide (LPS)-induced COX-2 protein expression in RAW246.7 cells[4]

Biochem/physiol Actions

Meloxicam, a member of the oxicam family, is a non-steroidal anti-inflammatory drug (NSAID). It is synthesized from piroxicam with aryl and the pyridyl ring substitution.[5] Meloxicam is a cyclooxygenase-2 (COX-2) inhibitor, it may be useful in treating inflammation and pain.[6] Meloxicam has been shown to stop thymulin-induced increas in concentration of cytokines and Nerve growth factor (NGF) in the liver of rats.[7] Meloxicam also suppresses tumor growth and may be capable of antagonizing cachexia in vitro.[8] It promotes apoptosis in hepatocellular carcinoma.[1]

Features and Benefits

This compound was developed by Boehringer Ingelheim. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

related product

Product No.
Description
Pricing

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of the specific cyclooxygenase-2 inhibitor meloxicam on tumour growth and cachexia in a murine model
Hussey HJ and Tisdale MJ
International Journal of Molecular Medicine, 87(1), 95-100 (2000)
The role of cytokines and prostaglandin-E2 in thymulin induced hyperalgesia
Safieh-Garabedian B, et al.
Neuropharmacology, 39(9), 1653-1661 (2000)
B Safieh-Garabedian et al.
Neuropharmacology, 39(9), 1653-1661 (2000-06-16)
We have recently reported that intraperitoneal (i.p.) injection of thymulin at low doses (50 ng) resulted in thermal and mechanical hyperalgesia and upregulation of the level of interleukin-1beta in the liver. In this study, we demonstrate that such injections of
Anti-Inflammatory Effect of (7R, 8S)-Dehydrodiconiferyl Alcohol-9? Gamma-Methyl Ether from the Rhizome of Belamcanda Chinensis: Role of Mir-146a and Mir-155
Binh BT, et al.
Biomedicine and Pharmacotherapy, 9(3), 909-918 (2016)
Cyclooxygenase-2 enzyme inhibitors: place in therapy.
Noble SL, et al.
Am. Fam. Physician., 61(12), 3669-3676 (2000)

Articles

Discover Bioactive Small Molecules for Lipid Signaling Research

Discover Bioactive Small Molecules for Lipid Signaling Research

Discover Bioactive Small Molecules for Lipid Signaling Research

Discover Bioactive Small Molecules for Lipid Signaling Research

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service