Skip to Content
Merck
All Photos(1)

Key Documents

D9885

Sigma-Aldrich

DEAE-Dextran hydrochloride

powder

Synonym(s):

Diethylaminoethyl-dextran hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25

biological source

bacterial (Leuconostoc mesenteroides)

form

powder

extent of labeling

~3.2% nitrogen

useful pH range

4.0 - 6.0

solubility

H2O: soluble 100 mg/mL, clear, colorless to faintly yellow

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

Application

DEAE-Dextran 500, a polycationic stabilizing molecule and transfection reagent, is used in the development of sustained release protein delivery and nucleic acid transfection systems. DEAE-Dextran 500 may be used for cell immobilization in biosensors.

Other Notes

To gain a comprehensive understanding of our extensive range of Dextrans for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Transfection using DEAE-dextran.
Selden RF.
Current Protocols in Immunology, 10, 10-10 (2001)
DEAE-dextran transfection of mammalian cultured cells.
E T Schenborn et al.
Methods in molecular biology (Clifton, N.J.), 130, 147-153 (1999-12-10)
Bhawna Yadav et al.
Cell surface (Amsterdam, Netherlands), 6, 100042-100042 (2020-12-29)
We designed experiments to assess whether fungal cell wall mannans function as an immune shield or an immune agonist. Fungal cell wall β-(1,3)-glucan normally plays a major and dominant role in immune activation. The outer mannan layer has been variously
Anita Bogdanov et al.
Antimicrobial agents and chemotherapy, 58(1), 405-413 (2013-11-06)
Chlamydiae are obligate intracellular bacteria that propagate in the inclusion, a specific niche inside the host cell. The standard method for counting chlamydiae is immunofluorescent staining and manual counting of chlamydial inclusions. High- or medium-throughput estimation of the reduction in
J Zarate et al.
Journal of microencapsulation, 28(7), 614-620 (2011-07-12)
Bovine serum albumin (BSA) loaded calcium alginate microparticles (MPs) produced in this study by a w/o emulsification and external gelation method exhibited spherical and fairly smooth and porous morphology with 1.052 ± 0.057 µm modal particle size. The high permeability of the calcium

Related Content

Dextrans are polysaccharides with molecular weights ≥1,000 Dalton, featuring a linear backbone of α-linked d-glucopyranosyl repeating units.

Dextrans are polysaccharides with molecular weights ≥1,000 Dalton, featuring a linear backbone of α-linked d-glucopyranosyl repeating units.

Dextrans are polysaccharides with molecular weights ≥1,000 Dalton, featuring a linear backbone of α-linked d-glucopyranosyl repeating units.

Dextrans are polysaccharides with molecular weights ≥1,000 Dalton, featuring a linear backbone of α-linked d-glucopyranosyl repeating units.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service