Skip to Content
Merck
All Photos(2)

Key Documents

P3369

Sigma-Aldrich

Protamine sulfate

solid, 90.0-110.0% dry basis,≥92% (chromatography), suitable for transduction

Synonym(s):

Salmine sulfate from salmon sperm, Protamine sulfate

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

Protamine sulfate, meets USP testing specifications

Agency

meets USP testing specifications

Assay

≥92% (chromatography)
90.0-110.0% dry basis

form

solid

mol wt

Mr 5000-10000

technique(s)

transduction: suitable

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Protamine sulfate is a cationic polypeptide having a molecular weight of 5 kDa, isolated from salmon sperm.[1]

Application

Protamine sulfate has been used in the transduction of
  • lentiviruses into human embryonic 293T cells[2]
  • shRNA into mesenchymal stem cells/marrow stromal cells (MSC)[3]
  • lenti Sca1-OCT4 plasmid into human cord blood hematopoietic progenitor CD34+ cells[4]

Biochem/physiol Actions

Alkaline protamine binds to negatively charged heparin. Use of protamine during cardiovascular surgery sometime triggers anaphylactic reactions.[5]
Protamine facilitates direct transfection of microbial cells, and of human cells using cationic liposomes. Protamine effectively neutralizes the anticoagulation effects of heparin.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Moosung Lee et al.
eLife, 9 (2020-12-18)
The immunological synapse (IS) is a cell-cell junction between a T cell and a professional antigen-presenting cell. Since the IS formation is a critical step for the initiation of an antigen-specific immune response, various live-cell imaging techniques, most of which
shRNA-mediated decreases in c-Met levels affect the differentiation potential of human mesenchymal stem cells and reduce their capacity for tissue repair
Rosova I, et al.
Tissue Engineering: Part A, 16(8), 2627-2639 (2010)
Reprogramming of Human Cord Blood CD34+ Cells into Induced MSCs
Hematopoietic Differentiation of Human Pluripotent Stem Cells, 91-101 (2015)
Meagan R Rollins et al.
Current protocols in immunology, 129(1), e97-e97 (2020-05-21)
T lymphocytes are capable of specific recognition and elimination of target cells. Physiological antigen recognition is mediated by the T cell receptor (TCR), which is an alpha beta heterodimer comprising the products of randomly rearranged V, D, and J genes.
F L Sorgi et al.
Gene therapy, 4(9), 961-968 (1998-02-12)
A polycationic peptide, protamine sulfate, USP, has been shown to be able to condense plasmid DNA efficiently for delivery into several different types of cells in vitro by several different types of cationic liposomes. The monovalent cationic liposomal formulations (DC-Chol

Questions

  1. In what solution can this particular protamine sulfate be dissolved? Sterile water, PBS, DMSO,...?

    1 answer
    1. This product is not tested for solubility. However, other Protamine sulfate products currently available are dissolved initially in 6N Hydrochloric acid at 25 mg/mL. This stock solution may then be diluted out with aqueous buffer or medium. In addition, a brief search of the literature reveals that this material may also be prepared by dissolving in HPLC grade water to a concentration of 10mg/ml. A stock solution was diluted to a final working concentration in growth media of 10?g/ml and stored at -20°C. This information has not been validated.
      Please see the link below to review this publication:
      http://eprints.nottingham.ac.uk/11280/1/PhD_Thesis.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service