Skip to Content
Merck
All Photos(1)

Documents

D3035

Sigma-Aldrich

Deoxyribonucleic acid from human placenta

buffered aqueous solution, sexed, female

Synonym(s):

DNA

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41106310
NACRES:
NA.52

biological source

human placenta

grade

for molecular biology

form

buffered aqueous solution

shipped in

dry ice

storage temp.

−20°C

InChI

1S/C15H31N3O13P2/c16-13-1-7(20)11(28-13)5-25-32(21,22)31-9-3-15(18)29-12(9)6-26-33(23,24)30-8-2-14(17)27-10(8)4-19/h7-15,19-20H,1-6,16-18H2,(H,21,22)(H,23,24)

InChI key

AWBASQCACWFTGD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Human placental DNA is isolated from a single female donor placenta, but will contain some maternal DNA. The sex has been determined using hybridization with DNA probes.

Application

Deoxyribonucleic acid from human placenta has been used:
  • to compare the efficiency of DNA quantification methods
  • as a standard in real-time PCR analysis of DNA samples from bladder cancer cell lines
  • as an internal control to estimate the degree of fragmentation of circulating DNA
  • for restriction digest analysis to identify low copy repeat regions of human chromosome 22q11
  • to compare the 70-bp P5 exon sequence between DNA of different human and primate species
  • to calculate copy number of genes, relative to normal human tissue
  • in PCR reactions
For use in Southern hybridizations.

Recommended products

Solutions of DNA have been stored successfully for several months at 4 C, in 10 mM Tris, pH 7.5 - 8.0, with 1 mM EDTA and without a bacteriostatic agent. At low concentrations, about 25 μg/ml, DNA tends to absorb onto the surfaces of plastic tubes.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alain R Thierry et al.
Nucleic acids research, 38(18), 6159-6175 (2010-05-25)
Although circulating DNA (ctDNA) could be an attractive tool for early cancer detection, diagnosis, prognosis, monitoring or prediction of response to therapies, knowledge on its origin, form and rate of release is poor and often contradictory. Here, we describe an
Joanne S Aveyard et al.
The Journal of molecular diagnostics : JMD, 6(4), 356-365 (2004-10-28)
Many tumors have large homozygous deletions of the CDKN2A locus (encoding p14(ARF) and p16) and of CDKN2B (p15). Our aim was to determine which gene is the major target in bladder cancer. We used quantitative real-time PCR (RTQ-PCR) to determine
Heather Hoover et al.
Journal of proteome research, 14(9), 3670-3679 (2015-07-08)
Tumor types can be defined cytologically by their regions of chromosomal amplification, which often results in the high expression of both mRNA and proteins of certain genes contained within the amplicon. An important strategy for defining therapeutically relevant targets in
Claire E L Smith et al.
PloS one, 12(9), e0185678-e0185678 (2017-09-29)
The imprinted gene PLAGL1 is an important regulator of apoptosis and cell cycle arrest. Loss of its expression has been implicated in tumorigenesis in a range of different cancers, and overexpression during fetal development causes transient neonatal diabetes mellitus (TNDM).
J E Collins et al.
Genome research, 7(5), 522-531 (1997-05-01)
A clone map consisting of YACs, cosmids, and fosmids has been constructed covering low copy repeat regions of human chromosome 22q11. A combination of clone restriction digest analysis, single-copy landmark content analysis, HindIII-Sau3AI fingerprinting, and sequencing of PCR products derived

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service