Skip to Content
Merck
All Photos(1)

Documents

C7594

Sigma-Aldrich

Casein Blocking Buffer

for Northern and Southern blotting, powder blend

Synonym(s):

blocking buffer

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12161700
NACRES:
NA.25

grade

for molecular biology

form

powder blend

technique(s)

Southern blotting: suitable

pH

6.8-7.2(reconstituted per directions)

solubility

H2O: soluble, faintly yellow (dissolves in 5-10 minutes.)

storage temp.

room temp

General description

Casein is a milk-derived substance routinely used in blocking buffers for Western, Southern and Northern Blot procedures. Blocking agents are used to reduce non-specific binding of detection reagents to the membranes.

Application

Suitable as a blocking Buffer for Western, Southern and Northern Blot membranes. Blocked membranes can then be used in either chemiluminescent, colorimetric, or DIG detection assays.

Features and Benefits

  • Better blocking performance than dried milk or BSA
  • Compatible with neutral and positively-charged membranes
  • Compatible with chemiluminescent, colorimetric , and DIG detection assays

Preparation Note

Dissolve entire contents of bottle in 800 mL molecular biology grade water. Stir until well mixed, then add water to final volume of 1L. Store at 2-8°C for up to one week.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ochratoxin A in some French wines: application of a direct competitive ELISA based on an OTA-HRP conjugate.
Radoi, A., et al.
Analytical Letters, 42, 1187-1187 (2009)
Serapion Pyrpassopoulos et al.
Methods in molecular biology (Clifton, N.J.), 1046, 389-403 (2013-07-23)
Many proteins that reside in the cytoplasm bind directly to cell membranes and play roles in signaling, adhesion, metabolism, cell structure, and cell motility. Several of these membrane-binding proteins, especially cytoskeletal proteins, have mechanical functions that result in the transmission
Tadatsura Koshika et al.
Immunology, 134(4), 386-397 (2011-11-03)
α1,3-Galactosyltransferase gene-knockout pigs transgenic for porcine cytotoxic T-lymphocyte antigen 4 immunoglobulin (pCTLA4-Ig) have been produced to reduce T-cell-mediated rejection following xenotransplantation. The level of soluble pCTLA4-Ig in their blood was greatly in excess of the therapeutic level in patients, rendering
Claire M Mahoney et al.
The Journal of biological chemistry, 284(13), 8898-8909 (2009-01-17)
The application of pulsed, low intensity ultrasound is emerging as a potent therapy for the treatment of complex bone fractures and tissue damage. Ultrasonic stimuli accelerate fracture healing by up to 40% and enhance tendon and ligament healing by promoting
Marlène Cassar et al.
Frontiers in neuroscience, 14, 232-232 (2020-04-16)
A hallmark feature of Alzheimer's disease (AD) and other Tauopathies, like Frontotemporal Dementia with Parkinsonism linked to chromosome 17 (FTDP-17), is the accumulation of neurofibrillary tangles composed of the microtubule-associated protein Tau. As in AD, symptoms of FTDP-17 include cognitive

Protocols

Protocol for Casein Blocking Buffer in Southern and Western blotting includes troubleshooting for optimizing signal.

Protocol for Casein Blocking Buffer in Southern and Western blotting includes troubleshooting for optimizing signal.

Protocol for Casein Blocking Buffer in Southern and Western blotting includes troubleshooting for optimizing signal.

Protocol for Casein Blocking Buffer in Southern and Western blotting includes troubleshooting for optimizing signal.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service