Skip to Content
Merck
All Photos(3)

Documents

M2015

Sigma-Aldrich

1-Stearoyl-rac-glycerol

≥99%

Synonym(s):

MG(18:0/0:0/0:0)[rac], 1-Monooctadecanoyl-rac-glycerol, rac-Glycerol 1-stearate, DL-α-Stearin, Monostearin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)16COOCH2CH(OH)CH2OH
CAS Number:
Molecular Weight:
358.56
Beilstein:
1728685
EC Number:
MDL number:
UNSPSC Code:
51191904
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic (organic)

Quality Level

Assay

≥99%

form

powder

functional group

phospholipid

lipid type

neutral glycerides

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCCCC(=O)OCC(O)CO

InChI

1S/C21H42O4/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-21(24)25-19-20(23)18-22/h20,22-23H,2-19H2,1H3

InChI key

VBICKXHEKHSIBG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Monostearin (1-Stearoyl-rac-glycerol) is used in the development of drug delivery vehicles such as nanoparticles and microemulsions.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Howida Kamal Ibrahim et al.
Drug delivery, 23(9), 3181-3192 (2016-03-05)
Statins proved potential bone healing properties. Rosuvastatin is a synthetic, hydrophilic, potent and highly efficacious statin. In the current work, an attempt was investigated to develop, evaluate various bioerodible composite sponges enclosing rosuvastatin and explore their potential in augmenting bone
Sagar S Bachhav et al.
International journal of pharmaceutics, 532(1), 612-622 (2017-09-25)
The oral uptake of intact nanocarriers through Peyer's patches is an important uptake pathway. We report Rifampicin Lipid-Polymer hybrid nanoparticles (RIF-LIPOMER) using glyceryl monostearate as lipid and the mucoadhesive polymer, Gantrez, with the objective of balancing hydrophobicity and mucoadhesion for
Maria Christina Camasmie Peters et al.
Journal of drug targeting, 28(3), 259-270 (2019-09-07)
Nanocrystals and lipid-based nanosystems have the potential to play a crucial role in a significant shift in the treatment of ophthalmic diseases. These drug delivery systems allow overcoming the barriers imposed by anatomy and physiology of the organ of vision.
Maria Chountoulesi et al.
Brain sciences, 10(6) (2020-06-06)
Multiple sclerosis (MS) is a chronic, autoimmune, neurodegenerative disease of the central nervous system (CNS) that yields to neuronal axon damage, demyelization, and paralysis. Although several drugs were designed for the treatment of MS, with some of them being approved
Mitali H Patel et al.
Drug development and industrial pharmacy, 45(8), 1242-1257 (2019-03-19)
Objective: The aim of the present investigation was to investigate the efficacy of solid lipid nanoparticles (SLNs) to enhance the absorption and bioavailability of lurasidone hydrochloride (LH) following oral administration. Methods: The LH loaded SLNs (LH-SLNs) were prepared by high

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service