Skip to Content
Merck
All Photos(2)

Key Documents

E0282

Sigma-Aldrich

MaxGel ECM

mixture, liquid

Synonym(s):

Human Basement Membrane Extract (BME)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
eCl@ss:
32190102
NACRES:
NA.71

Quality Level

form

liquid

storage temp.

−70°C

Looking for similar products? Visit Product Comparison Guide

Application

MaxGel ECM is used for the following applications:
  • Extracellular matrix (ECM) protein binding assays
  • Cell invasion assay
  • Cell culture
This product creates a three-dimensional environment that improves cellular proflieration by promoting cell growth and migration, and has been shown to aid the proliferation of many types of cells, including human embryonic stem cells, neural stem cells, neurons, glia, astrocytes, fibroblasts, hepatocytes, and keratinocytes.

Components

MaxGel ECM contains human extracellular matrix components including collagens, laminin, fibronectin, tenascin, elastin and a number of proteoglycans and glycosaminoglycans. This basement membrane extract reproduces the cooperative interactions between epithelia and mesenchyme that occur during the development and organotypic cell culture of skin.

Caution

MaxGel will remain stable for at least 1 year when stored at -70°C.

Legal Information

MaxGel is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H M Sharon Goh et al.
Applied and environmental microbiology, 79(21), 6535-6543 (2013-08-21)
Acinetobacter baumannii is a multidrug-resistant pathogen associated with hospital outbreaks of infection across the globe, particularly in the intensive care unit. The ability of A. baumannii to survive in the hospital environment for long periods is linked to antibiotic resistance
J Křivánek et al.
Folia biologica, 60 Suppl 1, 30-38 (2014-11-05)
Neural crest cells (NCCs) derive early in vertebrate ontogenesis from neural tube as a population of migratory cells with exquisite differentiation potential. Abnormalities in NCC behaviour are cause of debilitating diseases including cancers and a spectrum of neurocristopathies. Thanks to
Pyrimido[4,5-d]pyrimidin-4(1H)-one derivatives as selective inhibitors of EGFR threonine790 to methionine790 (T790M) mutants.
Tianfeng Xu et al.
Angewandte Chemie (International ed. in English), 52(32), 8387-8390 (2013-06-28)
Shi Song et al.
American journal of cancer research, 8(6), 993-1007 (2018-07-24)
Hepatocellular carcinoma (HCC) is one of the most prevalent and lethal cancers. It has been demonstrated that aberrant expression of miRNAs plays an important role in HCC development. Here, we observed decreased miR-125b expression status in tumor samples from HCC
Kang Li et al.
International journal of clinical and experimental pathology, 8(9), 10941-10946 (2015-12-01)
This study aims to explore the effects of vein subvolution combined with neural stem cells on nerve regeneration. Animal model of sciatic nerve defects was established with SD rats. A total of 63 SD rats were divided into control group

Articles

Human cell derived basement membrane extract (BME) hydrogel for 3D cell culture of primary cell, stem cell and xenograph cancer models.

3D cell culture overview. Learn about 2D vs 3D cell culture, advantages of 3D cell culture, and techniques available to develop 3D cell models

3D cell culture overview. Learn about 2D vs 3D cell culture, advantages of 3D cell culture, and techniques available to develop 3D cell models

3D cell culture overview. Learn about 2D vs 3D cell culture, advantages of 3D cell culture, and techniques available to develop 3D cell models

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service