Skip to Content
Merck
All Photos(2)

Key Documents

CLL1036

Sigma-Aldrich

U2OS GFP-HMGA1

human female bone (Disease source: Osteosarcoma)

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$585.00
5 G
$2,340.00

Vendor SKU: BB-0496-1G

$585.00


Ships in 7 days - from Aldrich Partner.

To order products, please contact your local dealer.

Select a Size

Change View
1 G
$585.00
5 G
$2,340.00

About This Item

UNSPSC Code:
41106514

biological source:
human female bone (Source Disease: Osteosarcoma)

Vendor SKU: BB-0496-1G

$585.00


Ships in 7 days - from Aldrich Partner.

To order products, please contact your local dealer.

Product Name

U2OS GFP-HMGA1,

biological source

human female bone (Source Disease: Osteosarcoma)

Quality Level

OMIM accession no.

storage temp.

−196°C

Gene Information

human ... HMGA1(3159)

General description

This product was derived from ATCC Cat. No. HTB-96.

U2OS GFP-HMGA1 are osteosarcoma, epithelial cells, from a human caucasian female (aged 15 years), with a GFP-tagged modification.

Application

This product is a human U2OS cell line in which the genomic HMGA1 gene has been endogenously tagged with a Green Fluorescent Protein (GFP) gene using CompoZr® Zinc Finger Nuclease technology. Integration resulted in endogenous expression of the fusion protein in which GFP is attached to the C-terminus of HMGA1. Fluorescence imaging shows characteristic HMGA1 nuclear expression. This stable cell line was expanded from a single clone. The target′s gene regulation and corresponding protein function are preserved in contrast to cell lines with overexpression via an exogenous promoter.
To learn more, please visit the Cellular Reporter Cell Line webpage

Features and Benefits

GFP-tagged on chromosome 6p21.

The U2OS cells are adherent, with a doubling time of approx. 29 hours.

Components

U2OS GFP-HMGA1 cells

Quality

Tested for Mycoplasma, sterility, post-freeze viability, short terminal repeat (STR) analysis for cell line identification, PCR assay for cell line species confirmation.

Preparation Note

Media Renewal changes two to three times per week.

Rapidly thaw vial by gentle agitation in 37°C water bath (~2 minutes), keeping vial cap out of the water. Decontaminate with 70% ethanol, add 9 mL culture media and centrifuge 125 x g (5-7 minutes). Resuspend in complete culture media and incubate at 37°C in a 5% CO2 atmosphere.

Subculture Ratio: approx. 1:3-1:6

The base medium for this cell line is McCoy′s 5A1 Medium Modified, Cat. No. M9309. To make the complete growth medium, add the following components to the base medium: fetal bovine serum, Cat. No. F4135, to a final concentration of 10%.

The cell freezing medium is DMSO 1X (Cat. No. C6164).

Legal Information

CompoZr is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service