Skip to Content
Merck
All Photos(2)

Key Documents

CLL1033

Sigma-Aldrich

U2OS GFP-LMNB1

human female bone (Disease source: Osteosarcoma)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
NACRES:
NA.81
biological source:
human female bone (Source Disease: Osteosarcoma)

Product Name

U2OS GFP-LMNB1,

biological source

human female bone (Source Disease: Osteosarcoma)

Quality Level

OMIM accession no.

storage temp.

−196°C

Gene Information

human ... lamin B1(4001)

General description

LMNB1 (lamin B1) is a B type lamin protein. LMNB1 is abundantly expressed in various cell types. This gene is located on human chromosome 5q23. The nuclear lamina, an intermediate filament meshwork, is made of LmnB1, LmnB2, A type (LmnA/C) and associated inner nuclear membrane proteins.
This product was derived from ATCC Cat. No. HTB-96.
U2OS GFP-LMNB1 are osteosarcoma, epithelial cells, from a human caucasian female (aged 15 years), with a GFP-tagged modification.

Application

This product is a human U2OS cell line in which the genomic LaminB1 gene has been endogenously tagged with a Green Fluorescent Protein (GFP) gene using CompoZr® Zinc Finger Nuclease technology. Integration resulted in endogenous expression of the fusion protein in which GFP is attached to the N-terminus of LaminB1. Fluorescence imaging shows the characteristic LaminB1 nuclear expression. This stable cell line was expanded from a single clone. The target′s gene regulation and corresponding protein function are preserved in contrast to cell lines with overexpression via an exogenous promoter.
To learn more, please visit the Cellular Reporter Cell Line webpage

Biochem/physiol Actions

Lamin B1 (LMNB1) is essential for proper chromosome condensation in interphase nuclei. It also participates in the distribution of heterochromatin and modulation of gene expression and splicing. Mutations in LMNB1 (lamin B1) gene results in the adult onset demyelinating disease autosomal dominant leukodystrophy (ADLD).
Nuclear lamin B1 encoded by LMNB1 gene is an intermediate filament protein of the nuclear envelope. The protein has transcriptional co-regulatory activity. It plays a key role in DNA replication, cellular aging and stress responses. Overexpression of the gene has been observed in hepatocellular carcinoma (HCC) patients. Thus, LMNB1 can be used as a potential biomarker for HCC.

Features and Benefits

GFP-tagged on chromosome 5q23.2
The U2OS cells are adherent, with a doubling time of approx. 29 hours.

Components

U2OS GFP-LMNB1 cells

Quality

Tested for Mycoplasma, sterility, post-freeze viability, short terminal repeat (STR) analysis for cell line identification, PCR assay for cell line species confirmation.

Preparation Note

Media Renewal changes two to three times per week.

Rapidly thaw vial by gentle agitation in 37°C water bath (~2 minutes), keeping vial cap out of the water. Decontaminate with 70% ethanol, add 9 mL culture media and centrifuge 125 x g (5-7 minutes). Resuspend in complete culture media and incubate at 37°C in a 5% CO2 atmosphere.

Subculture Ratio: approx. 1:3-1:6

The base medium for this cell line is McCoy′s 5A1 Medium Modified, Cat. No. M9309. To make the complete growth medium, add the following components to the base medium: fetal bovine serum, Cat. No. F4135, to a final concentration of 10%.

The cell freezing medium is DMSO 1X (Cat. No. C6164).

Legal Information

CompoZr is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

related product

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nuclear envelope rupture is induced by actin-based nucleus confinement.
Hatch EM and Hetzer MW
The Journal of Cell Biology, 215(1), 27?36-27?36 (2016)
The role of lamin B1 for the maintenance of nuclear structure and function.
Camps J, et al.
Nucleus (Austin, Tex.), 6(1), 8-14 (2015)
Defects of lipid synthesis are linked to the age-dependent demyelination caused by lamin B1 overexpression.
Rolyan H, et al.
The Journal of Neuroscience, 35(34), 12002-12017 (2015)
Circulating Lamin B1 (LMNB1) Biomarker Detects Early Stages of Liver Cancer in Patients
Sun S, et al.
Journal of Proteome Research, 9(1), 70-78 (2009)
Structural Organization of the Human Gene (LMNB1) Encoding Nuclear Lamin B1
Lin F, et al.
Genomics, 27(2), 230-236 (1995)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service