Skip to Content
Merck
All Photos(1)

Key Documents

SML3705

Sigma-Aldrich

Olaparib

≥98% (HPLC)

Synonym(s):

1-(Cyclopropylcarbonyl)-4-[5-[(3,4-dihydro-4-oxo-1-phthalazinyl)methyl]-2-fluorobenzoyl]piperazine, 4-[[3-[[4-(Cyclopropylcarbonyl)-1-piperazinyl]carbonyl]-4-fluorophenyl]methyl]-1(2H)-phthalazinone, AZD-2281, KU-0059436

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H23FN4O3
CAS Number:
Molecular Weight:
434.46
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

-10 to -25°C

SMILES string

Fc1c(cc(cc1)C=C4NNC(=O)c5c4cccc5)C(=O)N2CCN(CC2)C(=O)C3CC3

InChI

1S/C24H23FN4O3/c25-20-8-5-15(14-21-17-3-1-2-4-18(17)22(30)27-26-21)13-19(20)24(32)29-11-9-28(10-12-29)23(31)16-6-7-16/h1-5,8,13-14,16,26H,6-7,9-12H2,(H,27,30)

InChI key

JQYIJKFTPJWNFM-UHFFFAOYSA-N

Biochem/physiol Actions

Antineoplastic; Poly(ADP-ribose) polymerases-1 and 2 (PARP-1, PARP-2) inhibitor
Olaparib is an antineoplastic and potent inhibitor of poly(ADP-ribose) polymerases-1 and 2 (PARP-1, PARP-2) with an IC50 of 5 nM for PARP-1 and an IC50 of 1 nM for PARP-2. Olaparib is indicated to treat breast cancer, ovarian cancer, fallopian tube cancer, peritoneal cancer, pancreatic cancer, and prostate cancer.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Allosteric regulation of DNA binding and target residence time drive the cytotoxicity of phthalazinone-based PARP-1 inhibitors
Cell Chemical Biology, 29(12), 1694-1708 (2022)
Joana M Senra et al.
Molecular cancer therapeutics, 10(10), 1949-1958 (2011-10-06)
PARP-1 is a critical enzyme in the repair of DNA strand breaks. Inhibition of PARP-1 increases the effectiveness of radiation in killing tumor cells. However, although the mechanism(s) are well understood for these radiosensitizing effects in vitro, the underlying mechanism(s)
Evolution of poly(ADP-ribose) polymerase-1 (PARP-1) inhibitors. From concept to clinic.
Dana V Ferraris
Journal of medicinal chemistry, 53(12), 4561-4584 (2010-04-07)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service