Skip to Content
Merck
All Photos(1)

Key Documents

CAT100

Sigma-Aldrich

Catalase Assay Kit

sufficient for ≥100 tests enzymatic, determination of catalase activity in tissues and cells

Synonym(s):

Catalase Activity Detection Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

Quality Level

usage

sufficient for ≥100 tests enzymatic

detection method

colorimetric

shipped in

wet ice

storage temp.

2-8°C

Gene Information

human ... CAT(847)

General description

The Catalase Assay Kit contains all necessary components for studying catalase activity in various tissues and subcellular organelles.

Application

Sutitable for Colorimetric and UV Assays

Biochem/physiol Actions

Catalase is a ubiquitous antioxidant enzyme which catalyses the decomposition of hydrogen peroxide (H2O2) to water and oxygen. Hydrogen peroxide is formed in the eukaryotic cell as a by-product of various oxidases and superoxide dismutases. Hydrogen peroxide accumulation in cells causes oxidation of cellular targets such as DNA, proteins, and lipids leading to mutagenesis and cell death. Removal of the H2O2 from the cell by catalase provides protection against oxidative damage to living cells and its role in oxidative stress related diseases has been widely studied.

Features and Benefits

  • Useful for determining catalase activity - may be used in various tissues and cells
  • Simple, optimized protocol - A simple colorimetric assay for analysis of peroxisome enrichment and catalase activity

Suitability

Suitable for studying catalase activity in various tissues and subcellular organelles.

Principle

This assay method is based on the measurement of the hydrogen peroxide substrate remaining after the action of catalase. First, the catalase converts hydrogen peroxide to water and oxygen (catalatic pathway) and then this enzymatic reaction is stopped with sodium azide. An aliquot of the reaction mix is then assayed for the amount of hydrogen peroxide remaining by a colorimetric method.10 The colorimetric method uses a substituted phenol (3,5-dichloro-2-hydroxybenzenesulfonic acid), which couples oxidatively to 4-aminoantipyrine in the presence of hydrogen peroxide and horseradish peroxidase (HRP) to give a red quinoneimine dye (N-(4-antipyryl)-3-chloro-5-sulfonatep-benzoquinone-monoimine) that absorbs at 520 nm

Unit Definition

One unit of catalase will decompose 1.0 micromole of hydrogen peroxide to oxygen and water per minute at pH 7.0 at 25 °C at a substrate concentration of 50 mM hydrogen peroxide.

Preparation Note

Use ultrapure water in preparation of all solutions.

Kit Components Also Available Separately

Product No.
Description
SDS

  • P6782Peroxidase from horseradish, Type VI-A, essentially salt-free, lyophilized powder, ≥250 units/mg solid (using pyrogallol), 950-2000 units/mg solid (using ABTS) 5 mgSDS

  • 323381Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2OSDS

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Zámocký et al.
Progress in biophysics and molecular biology, 72(1), 19-66 (1999-08-14)
This review gives an overview about the structural organisation of different evolutionary lines of all enzymes capable of efficient dismutation of hydrogen peroxide. Major potential applications in biotechnology and clinical medicine justify further investigations. According to structural and functional similarities
M B Hampton et al.
FEBS letters, 414(3), 552-556 (1997-10-10)
The induction of apoptosis in Jurkat T-lymphocytes with 50 microM hydrogen peroxide was associated with caspase activation. Caspase activity was first detected 3 h after treatment, and the morphological features of apoptosis were apparent by 6 h. At higher concentrations
Andrea de Oliveira Falcão et al.
Food technology and biotechnology, 55(4), 553-561 (2018-03-16)
Enzymatic interesterification is used to manipulate oil and fat in order to obtain improved restructured lipids with desired technological properties. However, with raw materials containing significant amounts of bioactive compounds, the influence of this enzymatic process on the bioactivity of
S Tada-Oikawa et al.
FEBS letters, 442(1), 65-69 (1999-01-29)
Pulsed field gel electrophoresis showed that the initiation time of DNA breakage induced by the DNA alkylating agent duocarmycin A, which is not a redox-cycling agent, was almost the same in the human leukemia cell line HL-60 and its H2O2-resistant
M E Tome et al.
Cancer research, 61(6), 2766-2773 (2001-04-06)
Glucocorticoids are used for the treatment of lymphoid neoplasms, taking advantage of the well-known ability of these compounds to cause apoptosis in lymphoid tissues. Previously, we have shown that dexamethasone, a synthetic glucocorticoid, causes a down-regulation of several antioxidant defense

Articles

Centrifugation separates organelles based on size, shape, and density, facilitating subcellular fractionation across various samples.

Centrifugation separates organelles based on size, shape, and density, facilitating subcellular fractionation across various samples.

Centrifugation separates organelles based on size, shape, and density, facilitating subcellular fractionation across various samples.

Centrifugation separates organelles based on size, shape, and density, facilitating subcellular fractionation across various samples.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service