CS0009
Superoxide Dismutase, SOD, Activity Assay Kit
sufficient for 500 colorimetric tests
Synonym(s):
CU/ZN-SOD, SOD, SOD1, Superoxide Dismutase, Superoxide Dismutase 1 bovine, Superoxide: superoxide oxidoreductase
Sign Into View Organizational & Contract Pricing
All Photos(4)
About This Item
UNSPSC Code:
12161503
NACRES:
NA.54
Recommended Products
General description
Superoxide dismutase (SOD) is part of a family of metal ion-containing enzymes (SODs) that are found in most organisms, from bacteria to mammals. SODs catalyze the dismutation of superoxide (generated by aerobic respiration) to molecular oxygen and peroxide. Superoxide dismutase form the front line of defence against reactive oxygen species (ROS)-mediated injury and harbor anti-inflammatory activities. The Superoxide Dismutase (SOD) Activity Assay Kit provides a simple and sensitive procedure for measuring SOD enzymatic activity in various sample types. SOD activity is determined by measuring the decrease in superoxide anions (generated by the enzyme xanthine oxidase [XO]). Since superoxide anions interact with the provided WST dye, yielding color at 450 nm, the decrease in the color signal is proportional to SOD inhibition activity. The kit allows the determination of SOD activity in percent inhibition, or in units of activity using the provided SOD Enzyme as a standard. The provided SOD Enzyme can also be used to screen for SOD inhibitors.
The kit’s lowest limit of detection is 0.3 SOD units/mL. This kit can be used to measure SOD activity in biological samples such as serum, plasma and erythrocytes, tissue homogenates and cell lysates.
Components This kit contains sufficient reagents for 500 colorimetric tests.
Our economical Superoxide Dismutase (SOD) activity assay kit also saves your time and efforts:
The kit’s lowest limit of detection is 0.3 SOD units/mL. This kit can be used to measure SOD activity in biological samples such as serum, plasma and erythrocytes, tissue homogenates and cell lysates.
Components This kit contains sufficient reagents for 500 colorimetric tests.
Our economical Superoxide Dismutase (SOD) activity assay kit also saves your time and efforts:
- Highly sensitive, simple and high-throughput assay for measuring SOD activity in a variety of biological samples (serum, plasma,erythrocytes, tissue homogenates and cell lysates)
- Allows measurement of SOD activity both in percentage of inhibition and also by measuring the activity in enzymatic units using a calibration curve curve.
- Enables inhibitor screen using our SOD enzymes which is provided as a standard.
- User friendly: No need to weigh or calculate needed amounts of reagents on your own
- Supportivecalculator (Click here to download a calculator excel file):will calculate the reagents needed according to your experimental needs, andanalyses your results based on your experimental data!
- Quick instruction bench card - to assure your experimental success
- More experiments in one kit - contains sufficient reagents for 500 colorimetric tests.
- Limit of detection: 0.3 SOD units/mL
- Detection time: only 30 min
- Detection can be done at room temperature.
Features and Benefits
- Quantifying Superoxide Dismutase , SOD activity in biological samples
- Screening potential inhibitors for Superoxide Dismutase, SOD, activity
Suitability
This kit can be used to measure SOD activity in biological samples such as serum, plasma and erythrocytes, tissue homogenates and cell lysates.
Storage Class Code
10 - Combustible liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Alessia Remigante et al.
Frontiers in physiology, 14, 1225552-1225552 (2023-07-17)
Introduction: Aging is a process characterised by a decline in physiological functions. Reactive species play a crucial role in the aging rate. Due to the close relationship between aging and oxidative stress, functional foods rich in phytochemicals are excellent candidates
Dewa Ketut Meles et al.
Veterinary medicine international, 2022, 3685686-3685686 (2022-03-04)
2,3,7,8-tetrachlorodibenzo-p-dioxin (TCDD) is a persistent organic pollutant that induces overproduction of reactive oxygen species (ROS). Studies on avoiding the adverse effects of dioxin pollution exposure are needed in all aspects, including reproductive health. This study aimed to determine the effect
Masaki Kimura et al.
Cell, 185(22), 4216-4232 (2022-10-15)
Genotype-phenotype associations for common diseases are often compounded by pleiotropy and metabolic state. Here, we devised a pooled human organoid-panel of steatohepatitis to investigate the impact of metabolic status on genotype-phenotype association. En masse population-based phenotypic analysis under insulin insensitive
M Ardourel et al.
Experimental eye research, 225, 109282-109282 (2022-10-21)
Aging is a multifactorial process that affects the entire organism by cumulative alterations. Visual function impairments that go along with aging are commonly observed, causing lower visual acuity, lower contrast sensitivity, and impaired dark adaptation. Electroretinogram analysis revealed that the
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service