Skip to Content
Merck
All Photos(4)

Key Documents

C9322

Sigma-Aldrich

Catalase from bovine liver

lyophilized powder, 2,000-5,000 units/mg protein

Synonym(s):

H2O2:H2O2 oxidoreductase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
eCl@ss:
32160410
NACRES:
NA.54
Pricing and availability is not currently available.

biological source

bovine liver

Quality Level

form

lyophilized powder

specific activity

2,000-5,000 units/mg protein

mol wt

tetramer ~250 kDa

isoelectric point

5.4

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

SMILES string

O(CC)C(=O)c1ccc(cc1)O

InChI

1S/C9H10O3/c1-2-12-9(11)7-3-5-8(10)6-4-7/h3-6,10H,2H2,1H3

InChI key

NUVBSKCKDOMJSU-UHFFFAOYSA-N

Gene Information

cow ... CAT(280743)

Looking for similar products? Visit Product Comparison Guide

General description

Research Area: Cell Signaling

Catalase from bovine liver is a tetramer consisting of 4 equal subunits each with a 60 kDa molecular weight. Each of these subunits contains iron bound to a protoheme IX group. The enzyme will also strongly bind to NADP, where NADP and the heme group are within 13.7 angstroms.

Application

Catalase acts as a natural antioxidant to study the roles of reactive oxygen species in gene expression and apoptosis. It has also been used to protect against oxidative damage to proteins, lipids, and nucleic acids. Industrially, catalzes have been used to remove hydrogen peroxide added to milk and cheese, in textile bleaching, and to examine its positive effects on the viability of DNA-repair mutants of E. coli.

Catalase from bovine liver may be used:

  • to prepare H2O2-O2 based biocathode for applications in glucose biofuel cells
  • to study the kinetic properties and storage stability of catalase immobilized on to florisil
  • in glutathione-mediated superoxide generation in an aqueous solution

Biochem/physiol Actions

Catalase, an antioxidant enzyme found in all aerobic organisms, catalyzes the degradation of hydrogen peroxide, a byproduct of metabolic processes, into less harmful water and oxygen. It can also react with alkylhydrogen peroxides, such as methylperoxide and ethylperoxide and the second H2O2 molecule can be replaced by methanol, ethanol, propanol, formate and nitrate as a hydrogen donor. Catalase enzyme uses either iron (Fe) or manganese (Mn) as cofactor, and are classified as Fe-CAT or Mn-CAT.
This product doesn′t need any activators, but it is inhibited by 3-amino-1-H-1,2,4 triazole, cyanide, azide, hydroxylamine, cyanogens bromide, 2-mercaptoethanol, dithiothreitol, dianisidnie and nitrate.

Caution

Solutions of catalse should not be frozen. Frozen solution will result in a 50-70% loss of activity.

Unit Definition

One unit will decompose 1.0 μmole of H2O2 per min at pH 7.0 at 25 °C, while the H2O2 concentration falls from 10.3 to 9.2 mM, measured by the rate of decrease of A240.

Preparation Note

This product is a lyophilized powder with activity of 2,000-5,000 units/mg.
The enzyme is soluble in 50 mM potassium phosphate buffer at 1 mg/mL and pH 7.0.

Storage and Stability

Solutions of catalase should not be frozen. Freezing catalase solutions will lead to a loss of activity by 50-70%. The recommended storage temperature of the product in its powdered form is at -20 °C.

inhibitor

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christopher Gell et al.
Methods in cell biology, 95, 221-245 (2010-05-15)
In vitro assays that reconstitute the dynamic behavior of microtubules provide insight into the roles of microtubule-associated proteins (MAPs) in regulating the growth, shrinkage, and catastrophe of microtubules. The use of total internal reflection fluorescence microscopy with fluorescently labeled tubulin
Julie Elizabeth Keeble et al.
Pain, 141(1-2), 135-142 (2008-12-09)
Inflammatory diseases associated with pain are often difficult to treat in the clinic due to insufficient understanding of the nociceptive pathways involved. Recently, there has been considerable interest in the role of reactive oxygen species (ROS) in inflammatory disease, but
James C Dooley et al.
Current biology : CB, 30(12), 2404-2410 (2020-05-16)
Cortical development is an activity-dependent process [1-3]. Regarding the role of activity in the developing somatosensory cortex, one persistent debate concerns the importance of sensory feedback from self-generated movements. Specifically, recent studies claim that cortical activity is generated intrinsically, independent
Ahmet H Kaya et al.
Brain research, 1221, 93-97 (2008-06-21)
Serotonin (5-HT) containing neurons in the dorsal raphe nucleus (DRN) may play important roles in Parkinson's disease (PD). This study investigated neural and metabolic activity of the DRN in animal PD models based on dopamine depletion. The data show both
Amol Aher et al.
Developmental cell, 46(1), 40-58 (2018-06-26)
The dynamic instability of microtubules plays a key role in controlling their organization and function, but the cellular mechanisms regulating this process are poorly understood. Here, we show that cytoplasmic linker-associated proteins (CLASPs) suppress transitions from microtubule growth to shortening

Articles

Cellular oxidative stress is countered by enzymatic scavengers and antioxidant modulators against reactive oxygen species damage.

Cellular oxidative stress is countered by enzymatic scavengers and antioxidant modulators against reactive oxygen species damage.

Cellular oxidative stress is countered by enzymatic scavengers and antioxidant modulators against reactive oxygen species damage.

Cellular oxidative stress is countered by enzymatic scavengers and antioxidant modulators against reactive oxygen species damage.

Protocols

This procedure may be used for all Catalase products.

Questions

1–5 of 5 Questions  
  1. hi, because the catalase C100 is back ordered. we odered the powder of catalase, the catalog number is C9322. we would like to know how to make the powder into aqueous suspension and has the same concentration as catalase C100. thank you!

    1 answer
    1. The concentration of catalase C100 varies based on each lot. A sample Certificate of Analysis on the product page shows the mg/mL protein in suspension. Product C9322 is sold by mass, allowing calculation of the required mg/mL to achieve the same concentration. The enzyme is soluble in 50 mM potassium phosphate buffer at 1 mg/mL and pH 7.0. Both products also state their activity in Units/mg protein.

      Helpful?

  2. How can I reconstitute catalase and how can I store it in the refrigerator?

    1 answer
    1. A 10 mg/mL stock solution prepared in 50 mM potassium phosphate, pH 7.0, and 50% glycerol is stable for several months at 4°C, several weeks at room temperature, and at least 1 week in the freezer at -20°C. However, please note as mentioned in the "Caution" section of the Product Detail Page, freezing catalase solutions will lead to a loss of activity by 50-70%.

      Helpful?

  3. How can Catalase from bovine liver, Product C9322, be reconstituted?

    1 answer
    1. This product is soluble in 50 mM potassium phosphate, pH 7.0 at a concentration of 10 mg/mL.

      Helpful?

  4. What salts are present in Catalase from bovine liver, Product C9322?

    1 answer
    1. Catalase (C9322) contains potassium phosphate buffered salts (and 20% trehalose as a stabilizer).

      Helpful?

  5. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service