Skip to Content
Merck
All Photos(3)

Documents

C9142

Sigma-Aldrich

Cyanogen bromide-activated-Sepharose 4B

lyophilized powder

Synonym(s):

Cyanogen bromide-activated Agarose

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41106500
NACRES:
NA.56

form

lyophilized powder

technique(s)

affinity chromatography: suitable

matrix

Sepharose 4B

matrix active group

cyanate or related structures

matrix spacer

1 atom (when ligands are coupled via isourea derivative or related linkage)

swelling

1 g swells to 4-5 mL

particle size

45—165 μm

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Cyanogen bromide-activated-Sepharose is suitable for use in affinity chromatography and protein chromatography. It has been used to discover insecticidal agents, raise polyclonal antibodies against yeast invertase glycosyls, and study immunological processes of xenograft rejection.

C9142-15G′s updated product number is GE17-0430-01

Application

Cyanogen bromide-activated-Sepharose 4B has been used:
  • in the purification of clusterin from serum by an immunoaffinity-based separation procedure
  • in affinity chromatography to isolate Leishmania donovani protein antigens from the urine of visceral leishmaniasis patients
  • in affinity chromatography for the isolation of haptoglobin from human serum

Physical form

Lyophilized powder stabilized with lactose and dextran

Legal Information

Sepharose is a trademark of Cytiva

related product

Product No.
Description
Pricing

replaced by

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yan Ling et al.
The Journal of biological chemistry, 282(42), 30804-30816 (2007-08-29)
Farnesyl-diphosphate synthase (FPPS) catalyzes the synthesis of farnesyl diphosphate, an important precursor of sterols, dolichols, ubiquinones, and prenylated proteins. We report the cloning and characterization of two Toxoplasma gondii farnesyl-diphosphate synthase (TgFPPS) homologs. A single genetic locus produces two transcripts
Hans Gaus et al.
Biochemistry, 57(14), 2061-2064 (2018-03-29)
The Stabilin receptors are systemic clearance receptors for some classes of chemically modified nucleic acid therapeutics. In this study, the recombinant human secreted ecto-domain of the small isoform of Stabilin-2 (s190) was purified from cell culture and evaluated for direct
Dana A Massuto et al.
Reproduction (Cambridge, England), 139(2), 465-478 (2009-11-19)
The process of implantation is mediated by a complex network of signaling and adhesive factors. In the pig, latent and active transforming growth factor beta (TGFB), TGFB receptors (TGFBR), and integrins (ITGs) are present during the peri-implantation period. TGFB signals
Mark A Poli et al.
Journal of AOAC International, 90(1), 173-178 (2007-03-22)
A new competitive electrochemiluminescence-based immunoassay for the type-2 brevetoxins in oyster extracts was developed. The assay was verified by spiking known amounts of PbTx-3 into 80% methanol extracts of Gulf Coast oysters. We also provide preliminary data demonstrating that 100%
Qiong Zhao et al.
The Plant cell, 31(9), 2152-2168 (2019-06-22)
FYVE domain protein required for endosomal sorting1 (FREE1), a plant-specific endosomal sorting complex required for transport-I component, is essential for the biogenesis of multivesicular bodies (MVBs), vacuolar degradation of membrane protein, cargo vacuolar sorting, autophagic degradation, and vacuole biogenesis in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service