Skip to Content
Merck
All Photos(3)

Key Documents

T8512

Sigma-Aldrich

Activated Thiol–Sepharose 4B

lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41106500
NACRES:
NA.56

form

lyophilized powder

Quality Level

extent of labeling

1 μmol per mL

matrix

Sepharose 4B

matrix activation

cyanogen bromide

matrix active group

glutathione 2-pyridyl disulfide

matrix attachment

N-terminal amino group

matrix spacer

10 atoms (when ligands are coupled through the disulfide groups)

swelling

1 g swells to 4-5 mL

storage temp.

2-8°C

Application

Activated thiol Sepharose 4B is used in protein chromatography, affinity chromatography and activated/functionalized matrices. Activated thiol Sepharose 4B has been used to provide the first report of the isolation of aminopeptidase H from a reptile. Activated thiol Sepharose 4B has also been used to purify and characterize a neuropeptide-inactivating peptidase.

Physical form

Lyophilized powder stabilized with lactose and dextran

Legal Information

Sepharose is a trademark of Cytiva

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Oshima et al.
Biological & pharmaceutical bulletin, 23(5), 532-536 (2000-05-24)
Glutathione peroxidase (GPx) activity was detected in the ascite fluid of rats injected intraperitoneally with 2.5% heat-denatured casein solution. Activity in the ascite fluid increased with time after the injection of casein, and reached a maximum at 24 h. The
S Al-Jassabi
Biochemistry. Biokhimiia, 64(2), 217-222 (1999-04-03)
Aminopeptidase H was isolated and purified from fresh skeletal muscle of the lizard Agama stellio stellio by ammonium sulfate fractionation and successive chromatographies on DEAE-cellulose, Ultrogel AcA-34, activated thiol-Sepharose 4B, phenyl-Sepharose CL-4B, and DEAE-cellulose again. This is the first report
Abdullah Ozer et al.
Nucleic acids research, 41(14), 7167-7175 (2013-06-06)
The non-specific binding of undesired ligands to a target is the primary factor limiting the enrichment of tight-binding ligands in affinity selection. Solution-phase non-specific affinity is determined by the free-energy of ligand binding to a single target. However, the solid-phase
N Iwatsuki et al.
Biochemistry, 19(6), 1172-1176 (1980-03-18)
DNA photolyase purified from baker's yeast by affinity chromatography on UV-irradiated DNA noncovalently bound to cellulose and by chromatography on activated thiol-Sepharose 4B yields a single protein band having a molecular weight of 51 000 when analyzed by sodium dodecyl
N Agell et al.
The Biochemical journal, 273 ( Pt 3), 615-620 (1991-02-01)
A ubiquitin hydrolase that removes ubiquitin from a multi-ubiquitinated protein has been purified 600-fold from Saccharomyces cerevisiae. Four different ubiquitin-protein conjugates were assayed as substrates during the purification procedure. Enzymic activities that removed ubiquitin from ubiquitinated histone H2A, a ubiquitin-ubiquitin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service