Skip to Content
Merck
All Photos(2)

Key Documents

A2419

Sigma-Aldrich

ω-Aminohexyl–Sepharose 4B

aqueous ethanol suspension

Synonym(s):

ψ-Aminohexyl Resin, Aminohexyl Sepharose

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
23151817
NACRES:
NA.56
Pricing and availability is not currently available.

form

aqueous ethanol suspension

Quality Level

technique(s)

affinity chromatography: suitable

matrix activation

epoxy

matrix attachment

amino

matrix spacer

11 atoms

storage temp.

2-8°C

SMILES string

[X]NCCCCCCN

Application

ω-Aminohexyl– Sepharose 4B is used mainly in affinity chromatography. It has been used in mitochondrial research to purify thioredoxin reductase (TrxR2), which is important in cell signaling and may also be an anticancer drug target.

Physical form

Suspension in 20% ethanol.

Legal Information

Sepharose is a trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

95.0 °F

Flash Point(C)

35 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

M Burgos-Trinidad et al.
The Journal of biological chemistry, 267(5), 3498-3505 (1992-02-15)
The chick kidney mitochondrial cytochrome P-450 1,25-dihydroxyvitamin D3 24-hydroxylase was partially purified by sequential polyethylene glycol precipitation, aminohexyl-Sepharose 4B, and hydroxylapatite chromatography. The specific activity of the final preparation, when reconstituted with NADPH, adrenodoxin, and adrenodoxin reductase, was 245 pmol/min/mg
Sabine Urig et al.
Seminars in cancer biology, 16(6), 452-465 (2006-10-24)
Thioredoxin reductase (TrxR)-as part of a major thiol regulating system-allows redox metabolism to adjust to cellular requirements. Therefore, changes at the redox level reflect as a pars pro toto changes concerning the entire cell. Three different TrxR isoenzymes, TrxR1 as
J D Hayes et al.
The Biochemical journal, 207(3), 459-470 (1982-12-01)
Rat liver glutathione S-transferases have previously been defined by their elution behaviour from DEAE-cellulose and CM-cellulose as M, E, D, C, B, A and AA. These enzymes are dimeric proteins which comprise subunits of mol.wt. 22 000 (Ya), 23 500
F Pfennig et al.
The Journal of biological chemistry, 274(18), 12508-12516 (1999-04-23)
Actinomycin synthetase I (ACMS I) activates 4-methyl-3-hydroxyanthranilic acid, the precursor of the chromophoric moiety of the actinomycin, as adenylate. The gene acmA of ACMS I was identified upstream of the genes acmB and acmC encoding the two peptide synthetases ACMS
R Swanson et al.
The Journal of biological chemistry, 260(2), 1287-1289 (1985-01-25)
Bacterial luciferase from Vibrio harveyi, the 77,000-dalton light-emitting enzyme of the marine bacterium, has been crystallized into a two million cubic Angstrom cell with P212121 symmetry. The cell constants are a = 59.6 +/- 0.4 A, b = 112 +/-

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service