Skip to Content
Merck
All Photos(2)

Key Documents

C8147

Sigma-Aldrich

Hexadecyltrimethylammonium p-toluenesulfonate

Synonym(s):

Cetyltrimethylammonium p-toluenesulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)15N(CH3C6H4SO3)(CH3)3
CAS Number:
Molecular Weight:
455.74
Beilstein:
5470332
EC Number:
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
NACRES:
NA.25

description

cationic

Assay

≥98.0% (HPLC)

form

powder

mol wt

455.74 g/mol

SMILES string

Cc1ccc(cc1)S([O-])(=O)=O.CCCCCCCCCCCCCCCC[N+](C)(C)C

InChI

1S/C19H42N.C7H8O3S/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(2,3)4;1-6-2-4-7(5-3-6)11(8,9)10/h5-19H2,1-4H3;2-5H,1H3,(H,8,9,10)/q+1;/p-1

InChI key

MZMRZONIDDFOGF-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Hexadecyltrimethylammonium p-toluenesulfonate is a catatonic surfactant capable of forming wormlike micelles.

Application

Hexadecyltrimethylammonium p-toluenesulfonate has been used in a study to develop a silica grafted polydentate copper (II) complex. It has also been used in a study to investigate the dispersibility and bundle defoliation of single-walled carbon nanotubes.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sara B Lioi et al.
Physical chemistry chemical physics : PCCP, 11(41), 9315-9325 (2009-10-16)
Mixtures of oppositely charged surfactants, commonly called catanionic mixtures, are one of the most interesting and promising areas of colloidal chemistry. In this paper we review our previous work and report new results on electrostatic adsorption of organic solutes and
P H Purdy et al.
Cryobiology, 51(1), 102-112 (2005-07-05)
Cell plasma membrane fluidity is affected by membrane lipid and protein composition as well as temperature. Altering the cholesterol content of a membrane can change membrane fluidity at different temperatures and this may affect cell survival during cryopreservation. In these
N Tepale et al.
Journal of colloid and interface science, 363(2), 595-600 (2011-08-27)
The shear thickening behavior and the transition to shear thinning are examined in dilute cetyltrimethylammonium tosylate (CTAT) micellar solutions as a function of surfactant concentration and ionic strength using electrolytes with different counterion valence. Newtonian behavior at low shear rates
A Fischer et al.
Journal of colloid and interface science, 248(1), 163-168 (2005-11-18)
The sodium dodecyl benzene sulfonate (SDBS)/cetyltrimethyl-ammonium tosylate (CTAT)/water ternary system has previously been shown to give rise to the formation of vesicles for well-defined compositions requiring an excess of one of the surfactants over the other. Two types of vesicular
Lucia Zakharova et al.
Journal of colloid and interface science, 342(1), 119-127 (2009-11-07)
New amphiphilic pyrimidinic (AP) compounds with two ammonium head groups and different kinds of counterions, inorganic bromide anions (APB) and hydrophobic tosylate anions (APT) were synthesized. Self-organization in these systems has been studied by methods of tensiometry, conductometry, potentiometry and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service