Skip to Content
Merck
All Photos(1)

Documents

1102974

USP

Cetrimonium bromide

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Hexadecyltrimethylammonium bromide, CTAB, Cetrimonium bromide, Cetyltrimethylammonium bromide, Palmityltrimethylammonium bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)15N(Br)(CH3)3
CAS Number:
Molecular Weight:
364.45
Beilstein:
3598189
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

cetrimonium

manufacturer/tradename

USP

mp

248-251 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[Br-].CCCCCCCCCCCCCCCC[N+](C)(C)C

InChI

1S/C19H42N.BrH/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(2,3)4;/h5-19H2,1-4H3;1H/q+1;/p-1

InChI key

LZZYPRNAOMGNLH-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Cetrimonium bromide USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

related product

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2 - STOT RE 2 Oral - STOT SE 3

Target Organs

Gastrointestinal tract, Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hussaini Syed Sha Qhattal et al.
ACS nano, 8(6), 5423-5440 (2014-05-09)
Hyaluronan-grafted liposomes (HA-liposomes) preferentially target CD44-overexpressing tumor cells in vitro via receptor-mediated endocytosis. We investigated the pharmacokinetics and biodistribution of HA-liposomes with various sizes of HA (MW 5-8, 50-60, and 175-350 kDa) in mice. Incorporation of negatively charged HA on
Zhenzhi Shi et al.
Biomaterials, 35(25), 7058-7067 (2014-05-27)
Photodynamic therapy (PDT) is a promising treatment modality for cancer and other malignant diseases, however safety and efficacy improvements are required before it reaches its full potential and wider clinical use. Herein, we investigated a highly efficient and safe photodynamic
Lakshminarayana Polavarapu et al.
Small (Weinheim an der Bergstrasse, Germany), 10(15), 3065-3071 (2014-05-03)
The translation of a technology from the laboratory into the real world should meet the demand of economic viability and operational simplicity. Inspired by recent advances in conductive ink pens for electronic devices on paper, we present a "pen-on-paper" approach
Annemie Deiteren et al.
Gut, 63(12), 1873-1882 (2014-02-25)
Substantial evidence implicates mast cells and their main constituent histamine in the pathogenesis of visceral hypersensitivity. We explored the specific contribution of histamine H4 (H4R) and H1 (H1R) receptors to visceral hypersensitivity in a postinflammatory rat model. Trinitrobenzenesulfonic acid (TNBS)-colitis
Lan Ge et al.
American journal of physiology. Heart and circulatory physiology, 308(5), H500-H509 (2014-12-21)
Emerging evidence suggests a potential role of neutrophil extracellular traps (NETs) in linking sterile inflammation and thrombosis. We hypothesized that NETs would be induced during myocardial ischemia-reperfusion (I/R), and NET-mediated microthrombosis may contribute to myocardial "no-reflow". Male Wistar rats were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service