Skip to Content
Merck
All Photos(1)

Key Documents

B4136

Sigma-Aldrich

Benzyldimethylhexadecylammonium chloride

cationic detergent

Synonym(s):

16-BAC, Benzylhexadecyldimethylammonium chloride, Cetalkonium chloride, Cetyldimethylbenzylammonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)15N(Cl)(CH3)2CH2C6H5
CAS Number:
Molecular Weight:
396.09
Beilstein:
3802341
EC Number:
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
NACRES:
NA.25

Quality Level

form

powder

mol wt

396.09 g/mol

mp

55-65 °C (lit.)

storage temp.

room temp

SMILES string

[Cl-].CCCCCCCCCCCCCCCC[N+](C)(C)Cc1ccccc1

InChI

1S/C25H46N.ClH/c1-4-5-6-7-8-9-10-11-12-13-14-15-16-20-23-26(2,3)24-25-21-18-17-19-22-25;/h17-19,21-22H,4-16,20,23-24H2,1-3H3;1H/q+1;/p-1

InChI key

SXPWTBGAZSPLHA-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Benzyldimethylhexadecylammonium chloride is a cationic surfactant.

Application

Benzyldimethylhexadecylammonium chloride has been used in a study to assess the removal of cationic surfactants from aqueous solutions using activated carbon cloth (ACC). It has also been used in a study to investigate the prototropic behavior of the dye, 7-hydroxy-4-methylcoumarin (7H4MC).

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

143.6 °F - Pensky-Martens closed cup

Flash Point(C)

62 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sharmistha Dutta Choudhury et al.
The journal of physical chemistry. B, 113(19), 6736-6744 (2009-04-21)
The prototropic behavior of the dye, 7-hydroxy-4-methylcoumarin (7H4MC), has been studied in cationic benzyldimethylhexadecylammonium chloride (BDHC) and nonionic poly(oxyethylene)(tetramethylbutyl)phenyl ether (TritonX-100, TX-100) reverse micelles using ground-state absorption and steady-state and time-resolved fluorescence measurements. The results have been compared with the
Joe J Harrison et al.
Antimicrobial agents and chemotherapy, 52(8), 2870-2881 (2008-06-04)
Biofilms are slimy aggregates of microbes that are likely responsible for many chronic infections as well as for contamination of clinical and industrial environments. Pseudomonas aeruginosa is a prevalent hospital pathogen that is well known for its ability to form
J Sánchez-Martín et al.
Journal of hazardous materials, 174(1-3), 9-16 (2009-09-29)
Natural tannin-based adsorbent has been prepared on the basis of the gelification of Quebracho bark extract. The resulting product, Quebracho Tannin Gel (QTG) was tested as cationic dye adsorbent with Methylene Blue (MB). Kinetics of adsorption process were studied out
Zlata Grenoble et al.
The journal of physical chemistry. B, 117(1), 259-272 (2012-12-29)
The adsorption of the cationic surfactant benzyldimethylhexadecylammonium (BDMHA(+)) chloride has been studied at the hydrophilic silica-water interface by Raman spectroscopy in total internal reflection geometry (TIR Raman). This Raman spectroscopic technique takes advantage of an evanescent electric field that is
D E Macfarlane
Analytical biochemistry, 176(2), 457-463 (1989-02-01)
A two dimensional preparative polyacrylamide gel electrophoresis system is described which has both high resolving power and high loading capacity (approximately 100 mg protein). The first dimension is electrophoresis toward the cathode at acid pH in the presence of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service