Skip to Content
Merck
All Photos(3)

Key Documents

C8003

Sigma-Aldrich

5-α-Cholestane

≥97.0% (HPLC)

Synonym(s):

NSC 224419

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H48
CAS Number:
Molecular Weight:
372.67
Beilstein:
2051806
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥97.0% (HPLC)

form

powder

mp

80-82 °C (lit.)

SMILES string

CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC[C@H]4CCCC[C@]4(C)[C@H]3CC[C@]12C

InChI

1S/C27H48/c1-19(2)9-8-10-20(3)23-14-15-24-22-13-12-21-11-6-7-17-26(21,4)25(22)16-18-27(23,24)5/h19-25H,6-18H2,1-5H3/t20-,21-,22+,23-,24+,25+,26+,27-/m1/s1

InChI key

XIIAYQZJNBULGD-XWLABEFZSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

5-α-Cholestane has been used:
  • as an internal standard in gas phase chromatography (GC) and high-performance liquid chromatography (HPLC) to quantify cholesterol in meat products for a comparison study
  • to quantify the active compounds in the oils using a gas chromatograph coupled to a mass spectrometer
  • as a reference standard in GC analyses to quantify sterol content in the unsaponifiables obtained from oat lipids and different sterol standards like fucosterol, sitosterol, campesterol, and stigmasterol

Biochem/physiol Actions

5α-Cholestane is a sterol produced endogenously from cholesterol and has been isolated from human feces. It is derived from cholesterol by the action of intestinal microorganisms. Derivatives of 5α-cholestane in plants are called brassinosteroids that selectively activate the PPI3K/Akt pathway.

Preparation Note

5α-Cholestane yields clear to slightly hazy, colorless to faint yellow solution in chloroform at 50 mg/ml.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Abdulaziz A Alaqil et al.
Animals : an open access journal from MDPI, 10(9) (2020-09-10)
This study examines the effect of dietary supplementation with Lactobacillus acidophilus (LA) on the cholesterol levels, immune response, and productive performance of laying hens. A total of 216, 40-week-old, commercial Hy-Line brown chicken layers were randomly assigned into four treatment
Stefanie M Hixson et al.
PloS one, 11(3), e0152264-e0152264 (2016-03-25)
Nutritional enhancement of crops using genetic engineering can potentially affect herbivorous pests. Recently, oilseed crops have been genetically engineered to produce the long-chain omega-3 polyunsaturated fatty acids, eicosapentaenoic acid (EPA) and docosahexaenoic acid (DHA) at levels similar to that found
Comparative Analysis and Validation Methodologies of GC and HPLC for Analysis of Cholesterol in Meat Products
Stroher GL et al
American Journal of Analytical Chemistry, 3, 306-311 (2012)
Angelisa T Y Osmond et al.
Animals : an open access journal from MDPI, 11(4) (2021-05-01)
In this study, we evaluated whether oil extracted from the marine microbe, Schizochytrium sp. (strain T18), with high levels of docosahexaenoic acid (DHA), could replace fish oil (FO) in diets for rainbow trout (Oncorhynchus mykiss). Three experimental diets were tested:
L J Chen et al.
Journal of agricultural and food chemistry, 58(19), 10467-10474 (2010-09-11)
The objectives of this study were to develop a GC-MS method for determination of cholesterol oxidation products (COPs) in tea-leaf eggs and study the formation and inhibition of COPs as affected by heating time and various ingredients in marinated juice.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service