Skip to Content
Merck
All Photos(4)

Documents

C5157

Sigma-Aldrich

Campesterol

~65%

Synonym(s):

24α-Methyl-5-cholesten-3β-ol, 24(R)-Ergost-5-en-3β-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H48O
CAS Number:
Molecular Weight:
400.68
Beilstein:
3216975
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.28

biological source

Glycine max (soybean)

form

powder

concentration

~65%

shipped in

ambient

storage temp.

−20°C

SMILES string

[H][C@@]12CC=C3C[C@@H](O)CC[C@]3(C)[C@@]1([H])CC[C@]4(C)[C@H](CC[C@@]24[H])[C@H](C)CC[C@H](C)C(C)C

InChI

1S/C28H48O/c1-18(2)19(3)7-8-20(4)24-11-12-25-23-10-9-21-17-22(29)13-15-27(21,5)26(23)14-16-28(24,25)6/h9,18-20,22-26,29H,7-8,10-17H2,1-6H3/t19-,20+,22-,23-,24+,25-,26-,27-,28+/m0/s1

InChI key

SGNBVLSWZMBQTH-ZRUUVFCLSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Campesterol was used as standard in GC and HPLC analysis of oil samples from plants.

Biochem/physiol Actions

Campesterol is a phytosterol, primarily found in nuts, fruits, legumes and seeds. Though an analogue of cholesterol, it is poorly absorbed in humans and competitively inhibits the absorption of cholesterol. Campesterol decreases the transcription of genes involved in cholesterol metabolism in hepatocytes and enterocytes and has positive impact in treatment of cardiovascular disease.
Plant sterol. May lower absorption of dietary cholesterol.

Quality

Appears ~98% by HPLC and GC, but has been shown by 13C-NMR to contain ~35% dihydrobrassicasterol (24β-methyl-5-cholesten-3β-ol; 24[S]-ergost-5-en-3β-ol).

Preparation Note

Campesterol yields clear, colorless to faint yellow solution in chloroform at 20 mg/ml.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kang Jiang et al.
Journal of the American Society for Mass Spectrometry, 30(9), 1700-1712 (2019-05-22)
Phytosterols and tocopherols are essential for plant biochemistry, and they possess beneficial health effects for humans. Evaluating the tandem mass spectrometric (MS/MS) behavior of phytosterols and tocopherols is needed for the development of a qualitative and quantitative method for these
Hannu Päivä et al.
Clinical pharmacology and therapeutics, 78(1), 60-68 (2005-07-09)
Myopathy, probably caused by 3-hydroxy-3-methylglutaryl-coenzyme A reductase inhibition in skeletal muscle, rarely occurs in patients taking statins. This study was designed to assess the effect of high-dose statin treatment on cholesterol and ubiquinone metabolism and mitochondrial function in human skeletal
Juan P Barrios-Ramos et al.
Journal of the science of food and agriculture, 92(11), 2349-2357 (2012-03-21)
The effect of functional foods alone or in combination (cocoa + soy + oats + fish oil) on hepatic damage in rats affected with metabolic syndrome was investigated. Rats that were given cocoa showed a decrease in the levels of
D Lütjohann et al.
Journal of lipid research, 36(8), 1763-1773 (1995-08-01)
Absorption of dietary cholesterol, campesterol, and sitosterol, cholesterol balance, and fecal excretion of plant sterols were determined in three unrelated patients with phytosterolemia and three healthy volunteers during constant intake of cholesterol and plant sterols using accurate gas-liquid chromatography-mass spectrometry
Myung-Hee Lee et al.
Journal of ginseng research, 41(3), 428-433 (2017-07-14)
In this study, the fermentation of ginseng seeds was hypothesized to produce useful physiologically-active substances, similar to that observed for fermented ginseng root. Ginseng seed was fermented using The ginseng seed was fermented inoculating 1% of each strain on sterilized

Protocols

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service