Skip to Content
Merck
All Photos(1)

Key Documents

C4186

Sigma-Aldrich

Cytochrome c partially acetylated from equine heart

lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

form

lyophilized powder

Quality Level

100
200

composition

Protein, ~90% E0.1%/550

extent of labeling

~60% acetylated lysine residues (based on ninhydrin assay)

solubility

H2O: soluble 10 mg/mL, clear, red to red-brown

storage temp.

−20°C

Application

The enzyme has been used for the measuring superoxide production using the superoxide dismutase-inhibitable (SOD-inhibitable) cytochrome c reduction assay.

Packaging

Package size based on protein content.

Physical form

Contains potassium phosphate buffer salts.

Preparation Note

Dissolves in water to form a clear, red-brown colored solution at 10 mg/mL concentration.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Heidi C O'Neill et al.
Antioxidants & redox signaling, 10(2), 277-285 (2007-12-27)
Lipoic acid (LA) and its reduced product dihydrolipoic acid (DHLA) are potent antioxidants with capacity to scavenge reactive oxygen species (ROS) and recycle endogenous antioxidants. LA may increase cellular glutathione (GSH), an antioxidant lacking in the lung's epithelial lining fluid
The use of acetylated ferricytochrome c for the detection of superoxide radicals produced in biological membranes.
A Azzi et al.
Biochemical and biophysical research communications, 65(2), 597-603 (1975-07-22)
Carsten Wunderlich et al.
Pulmonary pharmacology & therapeutics, 21(3), 507-515 (2008-01-30)
Recently generated caveolin-1 deficient mice (cav-1 ko) suffer from severe lung fibrosis with marked pulmonary hypertension and arterial hypoxemia and may therefore serve as an useful animal model of this devastating human disorder. Accumulating evidence strongly supports the negative regulatory
Maria Simitsopoulou et al.
Methods in molecular medicine, 118, 161-179 (2005-05-13)
Candida albicans and non-albicans Candida spp. can cause serious infections in hospitalized and immunocompromised patients. A large number of antifungal agents and immunomodulators have been developed and may interact with both polymorphonuclear and mononuclear phagocytes against blastoconidia and pseudohyphae of
Ulf Landmesser et al.
The Journal of clinical investigation, 111(8), 1201-1209 (2003-04-17)
Tetrahydrobiopterin is a critical cofactor for the NO synthases, and in its absence these enzymes become "uncoupled," producing reactive oxygen species (ROSs) rather than NO. In aortas of mice with deoxycorticosterone acetate-salt (DOCA-salt) hypertension, ROS production from NO synthase is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service