Skip to Content
Merck
All Photos(8)

Key Documents

C2506

Sigma-Aldrich

Cytochrome c from equine heart

≥95% (SDS-PAGE)

Synonym(s):

Cytochrome c from horse heart

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.61

biological source

horse heart

Quality Level

Assay

≥95% (SDS-PAGE)

form

powder

mol wt

12,384

technique(s)

cell based assay: suitable

solubility

H2O: soluble 10 mg/mL

suitability

suitable for molecular biology

UniProt accession no.

application(s)

cell analysis

storage temp.

−20°C

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Cytochrome c has been used:
  • in the reduction of ferricytochrome c by xanthine oxidase-generated superoxide radicals
  • as an analyte by the single droplet deposition method using MALDI mass spectra
  • as a terminal electron acceptor in the assay of standard cytochrome c reductase activity
  • in cytochrome c oxidase histochemistry
  • in the measurement of cytochrome C oxidase activity
  • as a component of phosphate buffer for the detection and evaluation of complex IV activity using blue-native gel electrophoresis (BN-PAGE)
  • for detection of extracellular superoxide anion (ECSA) in isolated kidney phagocytes
The specific sites and extent of oxidation in horse cytochrome c treated with H2O2 and UV were characterized. It was suggested that the state of these sites could be used as a biomarker for the oxidative environment in a cell.

Biochem/physiol Actions

Cytochrome c is primarily known as an electron-carrying mitochondrial protein. The transition of cytochrome c between the ferrous and ferric states within the cell makes it an efficient biological electron-transporter and it plays a vital role in cellular oxidations in both plants and animals. It is generally regarded as a universal catalyst of respiration, forming an essential electron-bridge between the respirable substrates and oxygen
Cytochrome c, released from the outer mitochondrial compartment into the cytosol aids in activation of caspase-9 and initiation of the apoptotic protease cascade. This process is facilitated through its interaction with apoptotic protease-activating factor-1 (APAF-1) and deoxyadenosine triphosphate (dATP).(3) Various studies suggest that measurement of cytochrome C level in serum of tumor patients can be considered as a potential marker for cell death in vivo.

Preparation Note

Prepared using TCA.
This product in solution should be dark red to reddish brown when dissolved in water at 10 mg/mL.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B C Hill
The Journal of biological chemistry, 266(4), 2219-2226 (1991-02-05)
The reaction of the electrostatic cytochrome c-cytochrome oxidase complex with oxygen is measured by transient absorption spectroscopy. The oxygen reaction is initiated by photolytic removal of CO from cytochrome oxidase, using a flash-pumped dye laser. The subsequent reaction of the
Vittorio Pandini et al.
The Journal of biological chemistry, 277(50), 48463-48471 (2002-10-09)
Toxoplasma gondii possesses an apicoplast-localized, plant-type ferredoxin-NADP(+) reductase. We have cloned a [2Fe-2S] ferredoxin from the same parasite to investigate the interplay of the two redox proteins. A detailed characterization of the two purified recombinant proteins, particularly as to their
A novel role of the mitochondrial permeability transition pore in (-)-gossypol-induced mitochondrial dysfunction
Warnsmann V, et al.
Mechanisms of Ageing and Development, 170, 45-58 (2018)
Enzyme-catalyzed free radical reactions with nicotinamide adenine nucleotides. II. Lactate dehydrogenase-catalyzed oxidation of reduced nicotinamide adenine dinucleotide by superoxide radicals generated by xanthine oxidase.
P C Chan et al.
The Journal of biological chemistry, 249(4), 1317-1319 (1974-02-25)
Rapid extracellular release of cytochrome c is specific for apoptosis and marks cell death in vivo
Renz A, et al.
Blood, 98(5), 1542-1548 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service