Skip to Content
Merck
All Photos(3)

Key Documents

C3279

Sigma-Aldrich

Copper(II) chloride dihydrate

powder, BioReagent, suitable for cell culture

Synonym(s):

Cupric chloride dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CuCl2 · 2 H2O
CAS Number:
Molecular Weight:
170.48
EC Number:
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.75
Pricing and availability is not currently available.

biological source

synthetic (inorganic)

Quality Level

product line

BioReagent

form

powder

technique(s)

cell culture | mammalian: suitable

solubility

water: 100 mg/mL, clear, blue

SMILES string

O.Cl[Cu]Cl

InChI

1S/2ClH.Cu.2H2O/h2*1H;;2*1H2/q;;+2;;/p-2

InChI key

MPTQRFCYZCXJFQ-UHFFFAOYSA-L

Application

Copper(II) chloride dihydrate has been used as a positive control in dispersion of zebrafish embryo.[1] It has also been used for the preparation of trace metals mixture.[2][3]

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Copper-containing mesoporous bioactive glass promotes angiogenesis in an in vivo zebrafish model
Romero-Sanchez LB, et al.
Acta Biomaterialia, 68, 272-285 (2018)
Man Long Kwok et al.
Metallomics : integrated biometal science, 12(5), 732-751 (2020-03-24)
All cells require Cu as a cofactor, but Cu2+ induces toxicity and oxidative damage. A strict system is thus needed to maintain Cu homeostasis. Using the ZFL zebrafish liver cell line as a model, we studied the cellular responses after
Soluble and Solid Iron Reduction Assays with Desulfitobacterium hafniense
Comensoli L, et al.
Bio-protocol, 8(ARTICLE), 340-340 (2018)
Man Long Kwok et al.
Toxicology reports, 7, 822-835 (2020-07-17)
Copper (Cu) and cadmium (Cd) are widely used in industrial activities, resulting in Cu and Cd contamination in aquatic systems worldwide. Although Cu plays an essential role in many biological functions, an excessive amount of the metal causes cytotoxicity. In
Jianping Li et al.
Arteriosclerosis, thrombosis, and vascular biology, 39(3), 513-522 (2019-02-01)
Objective- We aimed to assess whether exposure to higher levels of ambient air pollution impairs HDL (high-density lipoprotein) function and to elucidate the underlying biological mechanisms potentially involved. Approach and Results- In the Beijing AIRCHD study (Air Pollution and Cardiovascular

Articles

Importance and uses of copper in serum-free eukaryotic, including hybridoma and Chinese Hamster Ovary (CHO), cell cultures

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service