Skip to Content
Merck
All Photos(2)

Key Documents

459097

Sigma-Aldrich

Copper(II) chloride dihydrate

Copper(II) chloride dihydrate

99.999%

Synonym(s):

Cupric chloride dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CuCl2 · 2H2O
CAS Number:
Molecular Weight:
170.48
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.999%
form:
solid
Pricing and availability is not currently available.

vapor density

>1 (vs air)

Quality Level

Assay

99.999%

form

solid

impurities

≤15.0 ppm Trace Metal Analysis

mp

100 °C (dec.) (lit.)

application(s)

battery manufacturing

SMILES string

Cl[Cu]Cl.[H]O[H].[H]O[H]

InChI

1S/2ClH.Cu.2H2O/h2*1H;;2*1H2/q;;+2;;/p-2

InChI key

MPTQRFCYZCXJFQ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Copper chloride dehydrate refluxed in acetonitrile can chemoselectively hydrolyze semicarbazones to carbonyl compounds. [1] Copper chloride dehydrate may be used as a catalyst for efficiently cleaving t-butyldimethylsilyl (TBDMS).[2]

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Deprotection of t-butyldimethylsiloxy (TBDMS) protecting group with catalytic copper (II) chloride dihydrate.
Ping Z, et al.
Chinese Chemical Letters = Zhongguo Hua Xue Kuai Bao, 11(19), 753-756 (2000)
Regeneration of carbonyl compounds from semicarbazones by copper (II) chloride dihydrate.
Ram RN and Varsha K.
Tetrahedron Letters, 32(41), 5829-5832 (1991)
Sebastian Müller et al.
PloS one, 13(11), e0206764-e0206764 (2018-11-07)
The clinically approved drug metformin has been shown to selectively kill persister cancer cells through mechanisms that are not fully understood. To provide further mechanistic insights, we developed a drug surrogate that phenocopies metformin and can be labeled in situ
Kenji Funaki et al.
Organic letters, 14(24), 6186-6189 (2012-12-12)
Direct arylation of thiophenes and benzothiophenes with aryltrimethylsilanes was effectively catalyzed by PdCl(2)(MeCN)(2) in the presence of CuCl(2) as an oxidant. The reaction preferentially occurred at the β-position of both thiophenes and benzothiophenes.
Mónica João Barros Amorim et al.
Environmental pollution (Barking, Essex : 1987), 164, 164-168 (2012-03-01)
Environmental effects of copper nanoparticles are little studied in terrestrial ecosystems. In the present article, the toxicity of copper nanoparticles (Cu-NP) on the enchytraeid Enchytraeus albidus is compared to the toxicity of a copper-salt (CuCl(2)). The effect parameters studied were

Questions

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service