Skip to Content
Merck
All Photos(1)

Documents

222011

Sigma-Aldrich

Copper(II) chloride

97%

Synonym(s):

Cupric chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CuCl2
CAS Number:
Molecular Weight:
134.45
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

reaction suitability

core: copper
reagent type: catalyst

mp

620 °C (lit.)

density

3.386 g/mL at 25 °C (lit.)

SMILES string

Cl[Cu]Cl

InChI

1S/2ClH.Cu/h2*1H;/q;;+2/p-2

InChI key

ORTQZVOHEJQUHG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Copper(II) chloride may be used in the preparation of copper(II)-chitosan complexes with potential applications in biomedical devices as antibiotic-free antibacterial biomaterials due to their cytocompatibility and antibacterial property.
Effective catalyst for the tetrahydropyranylation of alcohols, using mild conditions and in high yields.
Used with palladium in a catalytic synthesis of 3-haloindoles via an annulation process.

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marcin H Kudzin et al.
Marine drugs, 18(12) (2020-12-30)
In recent years, due to an expansion of antibiotic-resistant microorganisms, there has been growing interest in biodegradable and antibacterial polymers that can be used in selected biomedical applications. The present work describes the synthesis of antimicrobial polylactide-copper alginate (PLA-ALG-Cu2+) composite
Kanthasamy Raagulan et al.
Nanomaterials (Basel, Switzerland), 10(10) (2020-10-25)
The cost-effective spray coated composite was successfully synthesis and characterized by scanning electron microscopy, X-ray photoelectron spectroscopy, Raman spectroscopy, and X-ray diffraction techniques. The one step synthetic strategy was used for the synthesis of nanoplates that have a crystalline nature.
Synthesis, 1841-1841 (2007)
Olivija Plohl et al.
Nanomaterials (Basel, Switzerland), 9(2) (2019-02-10)
Due to the extreme rise of sludge pollution with heavy metals (e.g. copper), the options for its disposal or treatment are decreasing. On the contrary, properly heavy metal-cleaned sludge can be used as an alternative sustainable energy and agriculture source.
Bhalerao, U.T. et al.
Synthetic Communications, 26, 3081-3081 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service