Skip to Content
Merck
All Photos(1)

Key Documents

C0246

Sigma-Aldrich

Cabergoline

≥98% (HPLC)

Synonym(s):

1-Ethyl-3-(3′-dimethylaminopropyl)-3-(6′-allylergoline-8′β-carbonyl)urea, 1-[(6-Allylergoline-8β-yl)carbonyl]-1-[3-(dimethylamino)-propyl]-3-ethylurea, Cabaser, Dostinex, FCE-21336

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H37N5O2
CAS Number:
Molecular Weight:
451.60
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D −63 to −74°, c = 0.5 in dichloromethane

solubility

DMSO: ≥10 mg/mL

originator

Johnson & Johnson

storage temp.

2-8°C

SMILES string

CCNC(=O)N(CCCN(C)C)C(=O)[C@@H]1C[C@H]2[C@@H](Cc3c[nH]c4cccc2c34)N(CC=C)C1

InChI

1S/C26H37N5O2/c1-5-11-30-17-19(25(32)31(26(33)27-6-2)13-8-12-29(3)4)14-21-20-9-7-10-22-24(20)18(16-28-22)15-23(21)30/h5,7,9-10,16,19,21,23,28H,1,6,8,11-15,17H2,2-4H3,(H,27,33)/t19-,21-,23-/m1/s1

InChI key

KORNTPPJEAJQIU-KJXAQDMKSA-N

Gene Information

human ... DRD2(1813)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Cabergoline, a lysergic acid amide derivative, is a potent dopamine D2 receptor agonist. It also acts on dopamine receptors in lactophilic hypothalamus cells to suppress prolactin production in the pituitary gland. It has been used for monotherapy of Parkinson′s disease in the early phase; combination therapy, together with levodopa and a decarboxylase inhibitor such as carbidopa, in progressive-phase Parkinson′s disease and adjunctive therapy of prolactin-producing pituitary gland tumors (microprolactinomas).
Dopamine D2-receptor agonist; Prolactin inhibitor; Antiparkinsonian

Features and Benefits

This compound was developed by Johnson & Johnson. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Lact.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Samuel Santos-Ribeiro et al.
Journal of assisted reproduction and genetics, 32(7), 1063-1068 (2015-05-20)
We report on the results of the whole-genome analysis performed in a patient who developed severe ovarian hyperstimulation syndrome (OHSS) following gonadotropin-releasing hormone (GnRH) agonist triggering in a "freeze-all" protocol. A 30-year-old patient with polycystic ovary syndrome who developed severe
Monique P Curran et al.
Drugs, 64(18), 2125-2141 (2004-09-03)
Cabergoline is a synthetic ergoline dopamine agonist with a high affinity for dopamine D2 receptors and a long elimination half-life. This agent provides continuous dopaminergic stimulation with once-daily administration. Adjuvant oral cabergoline is usually well tolerated and effective in controlling
Valeria M S Leitao et al.
Fertility and sterility, 101(3), 664-675 (2013-12-24)
To evaluate the efficacy and safety of using cabergoline for reducing the risk of ovarian hyperstimulation syndrome (OHSS). Systematic review and meta-analysis of randomized clinical trials (RCTs). Women submitted to controlled ovarian stimulation (COS) for assisted reproduction. Cabergoline. Fertility centers.
Patrick Petrossians et al.
Neuroendocrinology, 92 Suppl 1, 116-119 (2010-09-21)
Dopamine (DA) is a catecholamine with a wide range of functions and whose five subtype receptors are found in different organs where they exert a mainly inhibitory action. Since this action may also appear in a number of secretory tumors
Renata S Auriemma et al.
European journal of endocrinology, 169(3), 359-366 (2013-07-05)
Cabergoline (CAB) has been found to be associated with increased risk of cardiac valve regurgitation in Parkinson's disease, whereas several retrospective analyses failed to detect a similar relation in hyperprolactinemic patients. The current study aimed at investigating cardiac valve disease

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service