Skip to Content
Merck
All Photos(1)

Key Documents

R2530

Sigma-Aldrich

Ropinirole hydrochloride

powder, ≥98% (HPLC)

Synonym(s):

4-[2-(Dipropylamino)ethyl]-1,3-dihydro-2H-indol-2-one hydrochloride, SKF 101468 hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H24N2O · HCl
CAS Number:
Molecular Weight:
296.84
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

light yellow

solubility

H2O: >10 mg/mL

originator

GlaxoSmithKline

SMILES string

Cl.CCCN(CCC)CCc1cccc2NC(=O)Cc12

InChI

1S/C16H24N2O.ClH/c1-3-9-18(10-4-2)11-8-13-6-5-7-15-14(13)12-16(19)17-15;/h5-7H,3-4,8-12H2,1-2H3,(H,17,19);1H

InChI key

XDXHAEQXIBQUEZ-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

An agonist at the D2 and D3 dopamine receptor subtypes, binding with higher affinity to D3 than to D2 or D4. It has negligible effect on D1-receptors. It has medium in vitro affinity to opioid receptors. Ropinirole is said to have virtually no affinity to 5-HT1, 5-HT2, benzodiazepine, GABA, muscarinic, α1-, α2-, and β-adrenoreceptors. Used as antiparkinsonian drug.
Ropinirole hydrochloride is effectively used to treat early and late Parkinson′s disease.

Features and Benefits

This compound is featured on the Dopamine Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by GlaxoSmithKline. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Francisco J Zagmutt et al.
The International journal of neuroscience, 122(7), 345-353 (2012-02-07)
The comparative safety profiles of monotherapeutic treatments for Parkinson's disease (PD) can provide valuable therapeutic information. The objective of this study was to perform an indirect comparison of Adverse Events (AEs) and Dropout Rates (DRs) among clinical trials of pramipexole
N Hattori et al.
Journal of clinical pharmacy and therapeutics, 37(5), 571-577 (2012-03-07)
Ropinirole hydrochloride, a dopamine receptor agonist with a non-ergot alkaloid structure, is highly selective for the dopamine D(2) /D(3) receptors. This study was conducted to evaluate the steady-state pharmacokinetics, safety and efficacy after repeated oral administration of prolonged-release tablets of
Dopamine agonist withdrawal syndrome in a patient with restless legs syndrome.
Benjamin J Dorfman et al.
Parkinsonism & related disorders, 19(2), 269-270 (2012-07-04)
T Nissen et al.
Scottish medical journal, 57(4), 217-220 (2012-09-25)
The expected duration of initial antiparkinson monotherapy before the need for supplementation is not clearly defined for routine practice. The aim of this study was to define the length of L-dopa (L-3, 4-dihydrophenylalanine) and dopamine agonist monotherapy. The duration of
Sébastien Duband et al.
Journal of forensic and legal medicine, 19(7), 422-425 (2012-08-28)
Ropinirole, a specific non-ergoline dopamine D2-receptor agonist, belongs to the drugs applied in treatment of Parkinson's disease (PD) and restless legs syndrome (RLS) and acts as a D2, D3, and D4 dopamine receptor agonist with highest affinity for D3. Therapeutic

Articles

We offer many products related to dopamine receptors for your research needs.

We offer many products related to dopamine receptors for your research needs.

We offer many products related to dopamine receptors for your research needs.

We offer many products related to dopamine receptors for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service