Skip to Content
Merck
All Photos(1)

Key Documents

Y0000472

Cabergoline

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

1-Ethyl-3-(3′-dimethylaminopropyl)-3-(6′-allylergoline-8′β-carbonyl)urea, 1-[(6-Allylergoline-8β-yl)carbonyl]-1-[3-(dimethylamino)-propyl]-3-ethylurea, Cabaser, Dostinex, FCE-21336

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H37N5O2
CAS Number:
Molecular Weight:
451.60
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

cabergoline

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

CCNC(=O)N(CCCN(C)C)C(=O)[C@@H]1C[C@H]2[C@@H](Cc3c[nH]c4cccc2c34)N(CC=C)C1

InChI

1S/C26H37N5O2/c1-5-11-30-17-19(25(32)31(26(33)27-6-2)13-8-12-29(3)4)14-21-20-9-7-10-22-24(20)18(16-28-22)15-23(21)30/h5,7,9-10,16,19,21,23,28H,1,6,8,11-15,17H2,2-4H3,(H,27,33)/t19-,21-,23-/m1/s1

InChI key

KORNTPPJEAJQIU-KJXAQDMKSA-N

Gene Information

human ... DRD2(1813)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Cabergoline EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Cabergoline, a lysergic acid amide derivative, is a potent dopamine D2 receptor agonist. It also acts on dopamine receptors in lactophilic hypothalamus cells to suppress prolactin production in the pituitary gland. It has been used for monotherapy of Parkinson′s disease in the early phase; combination therapy, together with levodopa and a decarboxylase inhibitor such as carbidopa, in progressive-phase Parkinson′s disease and adjunctive therapy of prolactin-producing pituitary gland tumors (microprolactinomas).

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Lact.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Renata S Auriemma et al.
European journal of endocrinology, 169(3), 359-366 (2013-07-05)
Cabergoline (CAB) has been found to be associated with increased risk of cardiac valve regurgitation in Parkinson's disease, whereas several retrospective analyses failed to detect a similar relation in hyperprolactinemic patients. The current study aimed at investigating cardiac valve disease
F Bogazzi et al.
Journal of endocrinological investigation, 31(12), 1119-1123 (2009-02-28)
Dopamine agonists have been associated with increased risk of cardiac valve regurgitation in patients with Parkinson's disease. Whether these drugs might be harmful for patients with hyperprolactinemia is still unsettled. Occasional case reports and 7 studies on the relationship between
Monique P Curran et al.
Drugs, 64(18), 2125-2141 (2004-09-03)
Cabergoline is a synthetic ergoline dopamine agonist with a high affinity for dopamine D2 receptors and a long elimination half-life. This agent provides continuous dopaminergic stimulation with once-daily administration. Adjuvant oral cabergoline is usually well tolerated and effective in controlling
Valeria M S Leitao et al.
Fertility and sterility, 101(3), 664-675 (2013-12-24)
To evaluate the efficacy and safety of using cabergoline for reducing the risk of ovarian hyperstimulation syndrome (OHSS). Systematic review and meta-analysis of randomized clinical trials (RCTs). Women submitted to controlled ovarian stimulation (COS) for assisted reproduction. Cabergoline. Fertility centers.
Patrick Petrossians et al.
Neuroendocrinology, 92 Suppl 1, 116-119 (2010-09-21)
Dopamine (DA) is a catecholamine with a wide range of functions and whose five subtype receptors are found in different organs where they exert a mainly inhibitory action. Since this action may also appear in a number of secretory tumors

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service