Skip to Content
Merck
All Photos(5)

Key Documents

B0770

Sigma-Aldrich

Brilliant Blue G

≥80% purity (HPLC), powder

Synonym(s):

Acid blue 90, Coomassie Brilliant Blue G

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C47H48N3NaO7S2
CAS Number:
Molecular Weight:
854.02
Colour Index Number:
42655
Beilstein:
5230822
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47
Pricing and availability is not currently available.

Product Name

Brilliant Blue G, pure

Quality Level

Assay

≥80.0% (HPLC)

form

powder

technique(s)

microbe id | staining: suitable

color

dark violet

solubility

H2O: 1 mg/mL

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].CCOc1ccc(Nc2ccc(cc2)C(\c3ccc(cc3C)N(CC)Cc4cccc(c4)S([O-])(=O)=O)=C5\C=CC(\C=C5C)=[N+](\CC)Cc6cccc(c6)S([O-])(=O)=O)cc1

InChI

1S/C47H49N3O7S2.Na/c1-6-49(31-35-11-9-13-43(29-35)58(51,52)53)40-21-25-45(33(4)27-40)47(37-15-17-38(18-16-37)48-39-19-23-42(24-20-39)57-8-3)46-26-22-41(28-34(46)5)50(7-2)32-36-12-10-14-44(30-36)59(54,55)56;/h9-30H,6-8,31-32H2,1-5H3,(H2,51,52,53,54,55,56);/q;+1/p-1

InChI key

RWVGQQGBQSJDQV-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Brilliant Blue G has been used for determining the urine protein concentration.[1] It has been used for the protein band detection after SDS-PAGE (sodium dodecyl sulfate polyacrylamide gel electrophoresis).[2][3] It has been used as a P2X7R (purinergic receptor) antagonist.[4]

Biochem/physiol Actions

Brilliant Blue G is used for staining protein(s) in polyacrylamide gels.[5] It is also a P2X7 purinergic receptor antagonist. The receptor is mainly involved in neuroinflammatory events. In human SOD1G93A amyotrophic lateral sclerosis mouse, administration of brilliant blue G reduces body weight loss. However, it does not affect survival or motor coordination in transgenic mice.[6]
P2X7 purinergic receptor antagonist.

Legal Information

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P2X7 antagonism using Brilliant Blue G reduces body weight loss and prolongs survival in female SOD1G93A amyotrophic lateral sclerosis mice.
PeerJ, e3064-e3064 (2017)
Melatonin attenuates hypertension-induced renal injury partially through inhibiting oxidative stress in rats.
Qiao YF, et al.
Molecular Medicine Reports, 13, 21-21 (2016)
Neuronal Release of Cytokine IL-3 Triggered by Mechanosensitive Autostimulation of the P2X7 Receptor Is Neuroprotective.
Lim JC, et al.
Frontiers in Cellular Neuroscience, 10, 270-270 (2016)
MATE transport of the E. coli-derived genotoxin colibactin.
Mousa JJ, et al.
Nature microbiology, 1, 15009-15009 (2016)
COMPARATIVE EVALUATION OF SOMATIC PROTEINS OF THE NYMPHAL STAGE OF LINGUATULA SERRATA ISOLATED FROM GOATS AND CATTLE IN NORTHWEST IRAN.
Bulgarian Journal of Veterinary Medicine, 19, 242-246 (2016)

Protocols

Simplify your SDS-PAGE gel preparation with our pre-mixed solutions and protocols to achieve precise protein separation.

Simplify your SDS-PAGE gel preparation with our pre-mixed solutions and protocols to achieve precise protein separation.

Simplify your SDS-PAGE gel preparation with our pre-mixed solutions and protocols to achieve precise protein separation.

Simplify your SDS-PAGE gel preparation with our pre-mixed solutions and protocols to achieve precise protein separation.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service