Skip to Content
Merck
All Photos(1)

Key Documents

B6760

Sigma-Aldrich

N-Benzoyl-L-tyrosine p-nitroanilide

≥98.0% (TLC)

Synonym(s):

Benzoyl-L-tyrosine p-nitroanilide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H19N3O5
CAS Number:
Molecular Weight:
405.40
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26

Product Name

N-Benzoyl-L-tyrosine p-nitroanilide,

Assay

≥98.0% (TLC)

Quality Level

form

powder

color

white

mp

235-237 °C

storage temp.

−20°C

SMILES string

Oc1ccc(CC(NC(=O)c2ccccc2)C(=O)Nc3ccc(cc3)[N+]([O-])=O)cc1

InChI

1S/C22H19N3O5/c26-19-12-6-15(7-13-19)14-20(24-21(27)16-4-2-1-3-5-16)22(28)23-17-8-10-18(11-9-17)25(29)30/h1-13,20,26H,14H2,(H,23,28)(H,24,27)

InChI key

CJERUMAUMMIPRF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N-Benzoyl-L-tyrosine p-nitroanilide (BTPNA) is used as a substrate to identify, differentiate and characterize serine carboxypeptidase(s) and various proteases.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Proteases in egg, miracidium and adult of Fasciola gigantica. Characterization of serine and cysteine proteases from adult.
Mohamed SA, Fahmy AS, et al.
Comp. Biochem. Physiol., B: Comp. Biochem., 142, 192-200 (2005)
Bernardo Ramírez-Zavala et al.
International journal of food microbiology, 91(3), 245-252 (2004-02-27)
We purified a carboxypeptidase (CPY) from the yeast of Kluyveromyces marxianus. This enzyme was purified 170 times from a soluble extract of 100000 x g. Purification consisted in a fractionated precipitation with ammonium sulfate and two chromatographic steps consisting of
K S LIU et al.
The Analyst, 115(8), 1143-1144 (1990-08-01)
In assaying chymotrypsin inhibition by the soybean Bowman-Birk inhibitor, two sequences of mixing the reactants were tried: adding the substrate last (s-last test) or adding the enzyme last (e-last test). The inhibition values obtained from the s-last test were either
Caterina Temporini et al.
Journal of separation science, 30(17), 3069-3076 (2007-10-11)
The preparation and optimization of a new monolithic chymotrypsin bioreactor for online protein digestion is described. Silica monolithic supports have been activated with epoxide functionalities following an optimized in situ procedure and used for covalent immobilization of chymotrypsin in one-step
G Colebatch et al.
Insect biochemistry and molecular biology, 31(4-5), 415-423 (2001-02-27)
Protease activities in the secreted saliva, salivary glands and midgut of the green mirid, Creontiades dilutus, were investigated. The saliva and salivary glands had more protease activity than the midgut, but no differences in protease activity levels were detected between

Articles

Analytical Enzyme Chymotrypsin: Chymotrypsin is produced in the acinar cells of the pancreas as the inactive precursor, chymotrypsinogen.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service