Skip to Content
Merck
All Photos(2)

Key Documents

B5388

Sigma-Aldrich

Brilliant Cresyl Blue ALD

Dye content, ≥60%, certified by the Biological Stain Commission, powder

Synonym(s):

3-Amino-7-(diethylamino)-2-methylphenoxazin-5-ium (T-4)-tetrachlorozincate (2:1), Brilliant Blue C, Brilliant Cresyl blue

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C17H20N3OCl · 1/2ZnCl2
CAS Number:
Molecular Weight:
385.96
Colour Index Number:
51010
Beilstein:
5690713
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Product Name

Brilliant Cresyl Blue ALD, certified by the Biological Stain Commission

grade

certified by the Biological Stain Commission

Quality Level

form

powder

composition

Dye content, ≥60%

solubility

water: 1 mg/mL, blue to very deep blue

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Zn-2](Cl)(Cl)(Cl)Cl.N(CC)(CC)c4cc5[o+]c6c(nc5cc4)cc(c(c6)N)C.N(CC)(CC)c1cc2[o+]c3c(nc2cc1)cc(c(c3)N)C

InChI

1S/2C17H20N3O.4ClH.Zn/c2*1-4-20(5-2)12-6-7-14-16(9-12)21-17-10-13(18)11(3)8-15(17)19-14;;;;;/h2*6-10H,4-5,18H2,1-3H3;4*1H;/q2*+1;;;;;+2/p-4

InChI key

SIHASHFMDFXGSX-UHFFFAOYSA-J

Application

Brilliant Cresyl Blue ALD has been used for the selection of oocytes. It has also been used for the selection of cumulus-oocyte complexes (COCs).

Biochem/physiol Actions

Brilliant Cresyl Blue ALD is mainly used for the selection of oocytes. It helps in determining the activity of glucose-6-phosphate dehydrogenase (G6PD). This enzyme is generated in growing oocytes. However, oocytes which complete their growth have low levels of G6PD. Low G6PD activity results in blue coloration of the cytoplasm because brilliant cresyl blue is not reduced to a colourless compound. It is also used for the staining in reticulocytes, where the dye stains reticulin.

Suitability

Certified as a supravital stain for reticulocytes.

Other Notes

The structure of Brilliant Cresyl Blue ALD contains a 2-methyl modification on the phenoxazine core, while the structure of the historic Brilliant Cresyl Blue contains a 8-methyl modification on the phenoxazine core. The biological staining application is the same for both structures.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vitrification of immature and matured bovine oocytes: effect of brilliant cresyl blue selection and hyaluronan addition.
Animal Reproduction, 13 (2016)
Liqin Wang et al.
Journal of biomedicine & biotechnology, 2012, 161372-161372 (2012-06-08)
Sheep oocytes derived from the ovaries collected from the slaughterhouse are often used for research on in vitro embryo production, animal cloning, transgenesis, embryonic stem cells, and other embryo biotechnology aspects. Improving the in vitro culture efficiency of oocytes can
Developmental competence of prepubertal goat oocytes selected with brilliant cresyl blue and matured with cysteamine supplementation.
Rodriguez-Gonzalez E, et al.
Reproduction, Nutrition, Development, 43, 179-179 (2003)
Cheesbrough M
District Laboratory Practice in Tropical Countries (2006)
David Berthier et al.
PloS one, 10(5), e0126498-e0126498 (2015-05-09)
Animal African Trypanosomosis particularly affects cattle and dramatically impairs livestock development in sub-Saharan Africa. African Zebu (AFZ) or European taurine breeds usually die of the disease in the absence of treatment, whereas West African taurine breeds (AFT), considered trypanotolerant, are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service